Skip to Content
Merck
All Photos(4)

Documents

227676

Sigma-Aldrich

Nickel(II) sulfate hexahydrate

ACS reagent, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 6H2O
CAS Number:
Molecular Weight:
262.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Quality Level

vapor density

2.07 (vs air)

Assay

≥98%

form

crystals

reaction suitability

reagent type: catalyst
core: nickel

impurities

≤0.002% N compounds
≤0.005% insolubles

anion traces

chloride (Cl-): ≤0.001%

cation traces

Ca: ≤0.005%
Co: ≤0.002%
Cu: ≤0.005%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
Mn: ≤0.002%
Na: ≤0.05%

SMILES string

O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2

InChI key

RRIWRJBSCGCBID-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) sulfate hexahydrate is a hydrated nickel salt. It is used as a nickel precursor for the preparation of Ni-based catalysts.

Application

Nickel(II) sulfate hexahydrate (NiSO4 6H2O) can be used as a nickel precursor for the preparation of:     
  • Palladium-nickel bimetallic nanoparticles (Pd-Ni) which are used as a catalyst in the Buchwald-Hartwig amination reaction.
  • NiFe2O4 nanoparticles (NPs) applicable for the fabrication of a glucose biosensor.

It can also be used as a recyclable catalyst in:      
  • The synthesis of α-aminophosphonates via a one-pot three-component reaction of aromatic aldehydes, primary amines, and diethylphosphite.      
  • Trifluoromethylation of arylamine in the presence of Langlois′ reagent (CF3SO2Na) as CF3 source and potassium persulfate (K2S2O8) as initiator.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun-Hua Lu et al.
Mikrochimica acta, 186(4), 227-227 (2019-03-09)
Water-soluble nitrogen-doped carbon quantum dots (C-dots) were fabricated by microwave-induced decomposition of the precursor materials citric acid and N,N'-bis(2-aminoethyl)-1,2-ethanediamine. The C-dots were placed on portable paper strips with novel origami designs to simplified user operations. The intensity of the blue
Polymorphism of nickel sulfate hexahydrate.
Angel RJ and Finger LW.
Acta Crystallographica Section C, Crystal Structure Communications, 44(11), 1869-1873 (1988)
Amperometric glucose biosensor based on NiFe2O4 nanoparticles and chitosan.
Luo L, et al.
Sensors and Actuators B, Chemical, 145(1), 293-298 (2010)
Mechanism and kinetics of thermal decomposition of nickel (II) sulfate (VI) hexahydrate.
Tomaszewicz E and Kotfica M.
Journal of Thermal Analysis and Calorimetry, 77(1), 25-31 (2004)
Charlotte G Mortz et al.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology, 24(1), 84-87 (2013-01-22)
We have previously reported patch test reactivity to nickel sulphate in a cohort of unselected infants tested repeatedly at 3-18 months of age. A reproducible positive reaction at 12 and 18 months was selected as a sign of nickel sensitivity provided a

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service