Skip to Content
Merck
All Photos(2)

Key Documents

497274

Sigma-Aldrich

Cobalt(II) thiocyanate

99.9% trace metals basis

Synonym(s):

Bis(thiocyanic acid)cobalt(II) salt, Cobaltous thiocyanate, Isothiocyanic acid cobalt(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(SCN)2
CAS Number:
Molecular Weight:
175.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

impurities

≤1500.0 ppm Trace Metal Analysis

SMILES string

N#CS[Co]SC#N

InChI

1S/2CHNS.Co/c2*2-1-3;/h2*3H;/q;;+2/p-2

InChI key

INDBQWVYFLTCFF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhimin Luo et al.
Small (Weinheim an der Bergstrasse, Germany), 12(43), 5920-5926 (2016-09-07)
A novel 3D cobalt sulfide (CoS) nanoparticle-decorated nitrogen and sulfur co-doped reduced graphene oxide aerogel (NSGA), referred to as CoS/NSGA, is prepared via three sequential processes, i.e., freeze-drying, annealing, and sulfidization. The obtained CoS/NSGA exhibits excellent electrocatalytic performance in the
J Lemli et al.
Pharmaceutisch weekblad. Scientific edition, 5(4), 142-144 (1983-08-26)
A rapid and simple colorimetric method is presented for the estimation of organic bases in pharmaceutical preparations. Solid cobalt thiocyanate reagent is used. Many bases form blue coloured ion pairs soluble in dichloromethane.
N El-Kousy et al.
Journal of pharmaceutical and biomedical analysis, 20(4), 671-679 (2000-03-07)
Atomic absorption spectrometry (AAS) and colourimetric methods have been developed for the determination of pizotifen (I), ketotifen (II) and loratadine (III). The first method depends on the reaction of the three drugs (I); (II) and (III) with cobalt thiocyanate reagent
H S Lee et al.
Journal of pharmaceutical and biomedical analysis, 21(1), 59-63 (2000-03-04)
For the simultaneous determination of berberine and palmatine from Phellodendri Cortex, Coptidis Rhizoma and pharmaceuticals, a new narrowbore HPLC method was developed with a simple and selective sample clean-up using cobalt thiocyanate reagent. Samples were sonicated with 2% hydrochloric acid
M Khan et al.
Journal of inorganic biochemistry, 75(2), 79-83 (1999-08-18)
A spectrophotometric study of cobalt(II), nickel(II) and copper(II) thiocyanato complexes was carried out in methanol at 25 degrees C and at a constant ionic strength of 1 M. Under the experimental conditions, two mononuclear complexes are identified with each of

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service