Skip to Content
Merck
All Photos(4)

Key Documents

191906

Sigma-Aldrich

Hydroxypropyl cellulose

average Mw ~1,000,000, powder, 20 mesh particle size (99% through)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

description

biological oxygen demand (BOD) 14,000 ppm

Quality Level

form

powder

autoignition temp.

752 °F

mol wt

average Mw ~1,000,000

impurities

<5 wt. %

particle size

20 mesh (99% through)

pH

5.0-8.5

interfacial tension mineral oil

12.5 dyn/cm, 0.1 wt. % in H2O (vs. mineral oil)

viscosity

1,275-3,500 cP, 1 wt. % in H2O(25 °C, Brookfield, spindle #3) (30 rpm)(lit.)

solubility

H2O: insoluble (above 45 °C)
polar organic solvents: soluble

density

0.5 g/mL at 25 °C (lit.)

SMILES string

N(C)(C)c1cc(c(cc1)C(N)CC)C

InChI

1S/C12H20N2/c1-5-12(13)11-7-6-10(14(3)4)8-9(11)2/h6-8,12H,5,13H2,1-4H3

InChI key

RRHXDYJWVYFMKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Thermo-responsive aqueous foams: Hydroxypropyl cellulose has been studied for its properties as a green polymer that can alter its solubility with temperature changes, making it suitable for responsive foam applications (Braunschweig, 2019).
  • Interaction mechanism in aqueous solutions: A study on the interaction between hydroxypropyl cellulose and water in solutions, focusing on the importance of polymer chain length in determining the physical properties of the solutions (Martin-Pastor and Stoyanov, 2020).
  • Corrosion inhibition: Hydroxypropyl cellulose has shown effectiveness as a corrosion inhibitor for aluminium in acidic environments, highlighting its potential in material science and engineering applications (Nwanonenyi et al., 2019).
  • Orally disintegrating films: This application involves the formulation design of hydroxypropyl cellulose films, aiming to enhance the delivery and dissolution profile of pharmaceuticals, making it significant in drug delivery and pharmaceutical sciences (Takeuchi et al., 2018).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homendra Naorem et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 101, 67-73 (2012-10-27)
The formation constant of triiodide ions from iodine-iodide equilibrium in aqueous-organic solvent or polymer mixed media have been determined spectrophotometrically at three different temperatures 20, 30 and 40°C. The organic solvents chosen for the study are ethylene glycol, 2-methoxy ethanol
Catherine Boissier et al.
Journal of pharmaceutical sciences, 101(7), 2512-2522 (2012-04-28)
Nuclear magnetic resonance (NMR) cryoporometry and scanning electron microscopy (SEM) image analysis have been used to investigate the size and shape distribution of pores in pharmaceutical coatings. The coatings were made from a mixture of hydroxypropylcellulose (HPC) and ethylcellulose (EC).
Kassibla E Dempah et al.
AAPS PharmSciTech, 14(1), 19-28 (2012-11-28)
Solid-state NMR spectroscopy (SSNMR), coupled with powder X-ray diffraction (PXRD), was used to identify the physical forms of gabapentin in samples prepared by recrystallization, spray drying, dehydration, and milling. Four different crystalline forms of gabapentin were observed: form I, a
Noorullah Naqvi Mohammed et al.
AAPS PharmSciTech, 13(4), 1158-1169 (2012-09-11)
The objective of this research work was to evaluate Klucel™ hydroxypropylcellulose (HPC) EF and ELF polymers, for solubility enhancement as well as to address some of the disadvantages associated with solid dispersions. Ketoprofen (KPR), a Biopharmaceutics Classification System class II
Satomi Onoue et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 46(5), 492-499 (2012-04-14)
The present study was undertaken to develop a solid self-emulsifying drug delivery system of coenzyme Q(10) (CoQ(10)/s-SEDDS) with high photostability and oral bioavailability. The CoQ(10)/s-SEDDS was prepared by spray-drying an emulsion preconcentrate containing CoQ(10), medium-chain triglyceride, sucrose ester of fatty

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service