Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hydroxypropyl cellulose
Synonym(s):
Hydroxypropyl cellulose
CAS Number:
9004-64-2
Products
Brand
Sigma-Aldrich (4)
Supelco (1)
Manufacturer
pH Value
Physical Form
Format
Quality Segment
Application
Polymer Composition
Polymer Type
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (2)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
435007
Hydroxypropyl cellulose,
average M
w
~80,000, average M
n
~10,000, powder, 20 mesh particle size (99% through)
Expand
View Pricing
191884
Hydroxypropyl cellulose,
average M
w
~100,000, powder, 20 mesh particle size (99% through)
Expand
View Pricing
191892
Hydroxypropyl cellulose,
average M
w
~370,000, powder, 20 mesh particle size (99% through)
Expand
View Pricing
191906
Hydroxypropyl cellulose,
average M
w
~1,000,000, powder, 20 mesh particle size (99% through)
Expand
View Pricing
1329800
Hydroxypropyl cellulose,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
PHR1485
Hydroxypropyl cellulose,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
Y0001720
Hydroxypropyl cellulose,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing