Skip to Content
Merck
All Photos(1)

Key Documents

09368

Sigma-Aldrich

Hydroxyethyl-cellulose

Synonym(s):

2-Hydroxyethyl cellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

form

powder

Quality Level

color

white to faint beige

viscosity

~145 mPa.s, 1 % in H2O(20 °C)

storage temp.

room temp

SMILES string

O1C(C(C(C(C1CO)OC)O)OCCO)OC2C(OC(C(C2O)O)OC4C(OC(C(C4O)O)C)CO)COC3OC(C(C(C3O)O)OC)CO

InChI

1S/C29H52O21/c1-10-15(34)16(35)24(13(8-33)45-10)49-28-20(39)18(37)25(50-29-26(43-5-4-30)21(40)23(42-3)12(7-32)47-29)14(48-28)9-44-27-19(38)17(36)22(41-2)11(6-31)46-27/h10-40H,4-9H2,1-3H3

InChI key

CWSZBVAUYPTXTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydroxyethylcellulose is used as a gelling and thickening agent in the development of biostructures for the delivery of hydrophobic drugs. Hydroxyethylcellulose is used in the development of polymer networks and block copolymers useful in separation technology such as capillary electrophoresis and in biofilms and coatings.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Semalty et al.
Indian journal of pharmaceutical sciences, 72(5), 571-575 (2011-06-23)
Enalapril maleate is used in the treatment of hypertension and angina pectoris. It shows low bioavailability due to high hepatic first pass metabolism. Hence the present work was undertaken to formulate mucoadhesive buccal films of enalapril maleate with an objective
Temperature-responsive self-assembly of charged and unchargedhydroxyethylcellulose-graft-poly(N-isopropylacrylamide) copolymer in aqueous solution.
Phan HT, Zhu K, Kj?niksen AL, Nystrom B.
Coll. Polymer Sci., 289, 993-1003 (2011)
Dan Zhou et al.
Journal of separation science, 34(23), 3441-3450 (2011-11-01)
To stabilize electroosmotic flow (EOF) and suppress protein adsorption onto the silica capillary inner wall, a cationic hydroxyethylcellulose-graft-poly (poly(ethylene glycol) methyl ether methacrylate) (cat-HEC-g-PPEGMA) graft copolymer composed of cationic backbone and bottle brush-like side chains was synthesized for the first
Fuhu Cao et al.
Electrophoresis, 32(10), 1148-1155 (2011-04-19)
In this work, a novel graft copolymer, hydroxyethylcellulose-graft-poly(2-(dimethylamino)ethyl methacrylate) (HEC-g-PDMAEMA), used as physical coatings of the bare fused-silica capillaries, was synthesized by using ceric ammonium nitrate initiator in aqueous nitric acid solution. EOF measurement results showed that the synthesized HEC-g-PDMAEMA
Runmiao Yang et al.
Electrophoresis, 29(7), 1460-1466 (2008-04-04)
We present cationized hydroxyethylcellulose (cat-HEC) synthesized in our laboratory as a novel physically adsorbed coating for CE. This capillary coating is simple and easy to obtain as it only requires flushing the capillary with polymer aqueous solution. A comparative study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service