Skip to Content
Merck
All Photos(2)

Key Documents

182605

Sigma-Aldrich

Poly(9-vinylcarbazole)

average Mw ~1,100,000, powder

Synonym(s):

PVK

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

powder

Quality Level

mol wt

average Mw ~1,100,000

refractive index

n20/D 1.683

transition temp

Tg 220 °C

density

1.2 g/mL at 25 °C (lit.)

Orbital energy

HOMO 5.8 eV 
LUMO 2.2 eV 

OLED Device Performance

ITO/PEDOT:PSS/PVK/PFO:Ir(btpy)3 (5 wt%)/Ca

  • Color: red
  • Max. Luminance: 726.5 Cd/m2
  • Max. EQE: 2.32 %

ITO/PEDOT:PSS/PVK:FIr6 (10 wt%)/TPBI/CsF:Al
  • Color: blue
  • Max. Luminance: 13800 Cd/m2
  • Max. EQE: 1.98 %
  • Turn-On Voltage: 5.5 V

ITO/PEDOT:PSS/PVK:Ir(mppy)2 (6 wt%)/Bphen/LiF:Al
  • Color: green
  • Max. Luminance: 10000 Cd/m2
  • Max. EQE: 1.1 %
  • Turn-On Voltage: 3.8 V

SMILES string

[n]1(c2c(c3c1cccc3)cccc2)C=C

InChI

1S/C14H11N/c1-2-15-13-9-5-3-7-11(13)12-8-4-6-10-14(12)15/h2-10H,1H2

InChI key

KKFHAJHLJHVUDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(9-vinylcarbazole) (PVK) is a conductive polymer which is mainly used as a hole transporting medium at high efficiencies with low driving voltage. It can also be used as an anode for hole injection and can act as an effective charge transferring gate by co-doping it with organic dyes.

Application

PVK:graphene oxide composite can be used as a hybrid film which can be coated on silica substrate for the fabrication of organic field effect transistors (OFETs). It can also form a nano-composite film with carbon nanospheres (CNS) which can be potentially used in supercapacitors and sensor based applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel carbon nanospheres and poly (9-vinylcarbazole) composites: Synthesis, structural and photo-physical properties of films elaboration
Mbarek M, et al.
Materials Chemistry and Physics, 211(3), 399-405 (2018)
Blue electroluminescent diodes utilizing blends of poly (p-phenylphenylene vinylene) in poly (9-vinylcarbazole)
Zhang C, et al.
Synthetic Metals, 62(1), 35-40 (1994)
Single-layered light-emitting diodes possessing methoxy-modified pyrazoloquinoline dyes in poly-N-vinylcarbazole matrix
Gondek E, et al.
Journal of Luminescence, 128(3), 348-354 (2008)
Poly (9-vinylcarbazole)-graphene oxide composite field-effect transistors with enhanced mobility
Aleshin AN, et al.
Organic Electronics, 16(3), 186-194 (2015)
Takuro Midorikawa et al.
Journal of nanoscience and nanotechnology, 7(4-5), 1419-1426 (2007-04-25)
Photoinduced electron-transfer and hole-migration processes for newly prepared copolymers of C60 and N-vinylcarbazole (VCz) with the several compositions have been investigated using time-resolved fluorescence and absorption methods. Shortening of fluorescence lifetime of the C60 moiety in PVCz-C60 was observed, suggesting

Articles

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service