Skip to Content
Merck
All Photos(1)

Key Documents

571652

Sigma-Aldrich

Poly(9,9-di-n-octylfluorenyl-2,7-diyl)

light-emitting polymer

Synonym(s):

Fluorenyl polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H9(C29H40)nC8H9
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

solid

mol wt

Mw ≥20000

solubility

chloroform: soluble

λmax

365 nm

fluorescence

λex 374 nm; λem 417 nm in chloroform(lit.)

OLED Device Performance

ITO/PEDOT:PSS/PVK/PFO/Ca

  • Color: blue
  • Max. Luminance: 102.3 Cd/m2
  • Max. EQE: 0.17 %

ITO/PEDOT:PSS/PVK/PFO:Ir(btpy)3 (5 wt%)/Ca
  • Color: red
  • Max. Luminance: 726.5 Cd/m2
  • Max. EQE: 2.32 %

ITO/PEDOT:PSS/PVK/PFO:Iridium (III) tris(2-(4-tolyl)pyridinato-N,C2) (5 wt%)/Ca
  • Color: green
  • Max. Luminance: 3311 Cd/m2
  • Max. EQE: 2.31 %

Mw/Mn

~3.7

Looking for similar products? Visit Product Comparison Guide

General description

Poly(9,9-di-n-octylfluorenyl-2,7-diyl) (PFO) is a blue light emitting fluorescent polymer with high electroluminescence quantum yield and chemical stability. It can form an emissive layer on the surface of the optoelectronic devices.

Application

PFO can be used as a phase separated polymer that interacts with the conducting probes to define the effect of noise sources on σ-conjugated charge carriers. It is used to disperse single walled carbon nanotubes (SWCNTs), which can be further used in the fabrication of field-effect transistors (FETs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel electrochemiluminescent biosensor based on resonance energy transfer between poly (9, 9-di-n-octylfluorenyl-2, 7-diyl) and 3, 4, 9, 10-perylenetetracar-boxylic acid for insulin detection.
Zhang H, et al.
Biosensors And Bioelectronics, 104, 65-71 (2018)
High Performance Ambipolar Field-Effect Transistor of Random Network Carbon Nanotubes.
Bisri SZ, et al.
Advanced Materials, 24(46), 6147-6152 (2012)
Nanoscale direct mapping of localized and induced noise sources on conducting polymer films.
Shekhar S, et al.
Nanoscale, 8(2), 835-842 (2016)
Effectiveness of sorting single-walled carbon nanotubes by diameter using polyfluorene derivatives.
Gao, J, et al.
Carbon, 49(1), 333-338 (2011)
Mapping nanoscale effects of localized noise-source activities on photoconductive charge transports in polymer-blend films
Shekhar S, et al.
Nanotechnology, 29(20), 205204-205204 (2018)

Articles

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service