Skip to Content
Merck
All Photos(2)

Key Documents

368350

Sigma-Aldrich

Poly(9-vinylcarbazole)

average Mn 25,000-50,000

Synonym(s):

PVK

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

mol wt

average Mn 25,000-50,000

Quality Level

mp

>300 °C

density

1.2 g/mL at 25 °C (lit.)

Orbital energy

HOMO 5.8 eV 
LUMO 2.2 eV 

OLED Device Performance

ITO/PEDOT:PSS/PVK/PFO:Ir(btpy)3 (5 wt%)/Ca

  • Color: red
  • Max. Luminance: 726.5 Cd/m2
  • Max. EQE: 2.32 %

ITO/PEDOT:PSS/PVK:FIr6 (10 wt%)/TPBI/CsF:Al
  • Color: blue
  • Max. Luminance: 13800 Cd/m2
  • Max. EQE: 1.98 %
  • Turn-On Voltage: 5.5 V

ITO/PEDOT:PSS/PVK:Ir(mppy)2 (6 wt%)/Bphen/LiF:Al
  • Color: green
  • Max. Luminance: 10000 Cd/m2
  • Max. EQE: 1.1 %
  • Turn-On Voltage: 3.8 V

Mw/Mn

~2

SMILES string

[n]1(c2c(c3c1cccc3)cccc2)C=C

InChI

1S/C14H11N/c1-2-15-13-9-5-3-7-11(13)12-8-4-6-10-14(12)15/h2-10H,1H2

InChI key

KKFHAJHLJHVUDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(9-vinylcarbazole) (PVK) is a conjugated polymer with hydrophobic properties. PVK films have a glass transition temperature (Tg) of 200°C and are mainly used as hole transport materials for semiconductor application.

Application

PVK can be used for the synthesis of titanium oxide (TiO2) quantum dots fillers that find potential applications as donor-acceptor layers in polymeric solar cells. It may be used to functionalize carbon nanotubes (CNTs) and form conductive reinforcements on epoxy groups. Iridium composites may be surface modified by PVK for the fabrication of highly efficient organic light emitting devices.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microstructure evolution, thermal stability and fractal behavior of water vapor flow assisted in situ growth poly (vinylcarbazole)-titania quantum dots nanocomposites.
Mombru D, et al.
Journal of Physics and Chemistry of Solids, 111(9), 199-206 (2017)
Use of poly (9-vinylcarbazole) as host material for iridium complexes in high-efficiency organic light-emitting devices.
Yang M and Tsutsui T
Japanese Journal of Applied Physics, 39(8A), L828-L828 (2000)
Efficient white polymer light-emitting diodes from single polymer exciplex electroluminescence.
Liang J, et al.
Journal of Material Chemistry C, 5(9), 2397-2403 (2017)
Poly (9-vinylcarbazole) as a hole transport material for efficient and stable inverted planar heterojunction perovskite solar cells.
Yang L, et al.
Solar Energy Materials and Solar Cells, 163, 210-217 (2017)
Noncovalent functionalization of carbon nanotube using poly (vinylcarbazole)-based compatibilizer for reinforcement and conductivity improvement in epoxy composite.
Liu R, et al.
Journal of Applied Polymer Science, 134(26), 199-206 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service