178500
3-Methyl-1,2-cyclopentanedione
99%
Synonym(s):
2-Hydroxy-3-methyl-2-cyclopenten-1-one, Maple lactone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
Beilstein:
2076520
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
solid
mp
104-108 °C (lit.)
functional group
ketone
SMILES string
CC1CCC(=O)C1=O
InChI
1S/C6H8O2/c1-4-2-3-5(7)6(4)8/h4H,2-3H2,1H3
InChI key
OACYKCIZDVVNJL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
3-Methyl-1,2-cyclopentanedione is present in coffee and is capable of scavenging peroxynitrite. It also has anti-inflammatory effects.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
M Mozaffarieh et al.
Survey of ophthalmology, 53(5), 479-505 (2008-10-22)
Glaucomatous optic neuropathy implies loss of retinal ganglion cells, including their axons, and a major tissue remodeling, especially in the optic nerve head. Although increased intraocular pressure is a major risk factor for glaucomatous optic neuropathy, there is little doubt
Seo Young Choi et al.
Biochimica et biophysica acta, 1770(12), 1612-1619 (2007-10-13)
Peroxisome proliferator-activated receptors (PPARs) are members of the nuclear hormone receptor superfamily that plays a pivotal role in regulating inflammatory gene expression. The purpose of this study was to investigate the effects of coffee extract, 3-methyl-1,2-cyclopentanedione (3-MCP) on PPARs in
Bryson W Katona et al.
The Journal of organic chemistry, 72(24), 9298-9307 (2007-10-26)
Deoxycholic acid (DCA) is an endogenous secondary bile acid implicated in numerous pathological conditions including colon cancer formation and progression and cholestatic liver disease. DCA involvement in these disease processes results partly from its ability to modulate signaling cascades within
Jae Heun Chung et al.
Journal of agricultural and food chemistry, 55(16), 6787-6792 (2007-07-17)
Activation of the redox-sensitive transcription factor, nuclear factor-kappa B (NF-kappaB), plays a central role in inflammation and aging processes by inducing pro-inflammatory genes. The present study was designed to unravel the molecular mechanisms underlying the anti-inflammation effects of 3-methyl-1,2-cyclopentanedione (3-MCP)
Ae Ra Kim et al.
The Journal of pharmacy and pharmacology, 54(10), 1385-1392 (2002-10-25)
It has been known that reactive oxygen and nitrogen species such as nitric oxide (NO), superoxide radical (*O2-) and their byproduct peroxynitrite (ONOO-) induce cellular and tissue injury, ultimately resulting in several human diseases. In this study, we examined scavenging
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service