Skip to Content
Merck
All Photos(3)

Key Documents

C101605

Sigma-Aldrich

1,3-Cyclohexanedione

97%

Synonym(s):

Dihydroresorcinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H8(=O)2
CAS Number:
Molecular Weight:
112.13
Beilstein:
385888
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

powder

contains

1-3% sodium chloride as stabilizer

mp

101-105 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1CCCC(=O)C1

InChI

1S/C6H8O2/c7-5-2-1-3-6(8)4-5/h1-4H2

InChI key

HJSLFCCWAKVHIW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Cyclohexanedione can be used as a building block in the synthesis of:
  • 9-substituted 1,8-dioxo-xanthenes by reacting with unactivated aldehydes via aldol condensation/ Michael addition reaction.[1]
  • [(1,2,3-triazol-4-yl)methoxy-phenyl]-2H-indazolo[2,1-b]phthalazine-trione derivatives by treating with phthalhydrazide, aromatic propargyloxy aldehydes, and azides via a four-component condensation reaction.[2]
It can be also used to prepare 2-substituted adducts, which are important intermediates for the synthesis of ß-enaminones [3], polyhydroquinoline derivatives [4], 1,8-dioxo-dodecahydroxanthenes [5], spirocyclopentanols [6], oxathioles [7], and triquinanes.[8]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkylation of Cyclic 1,3-Diketones
Thennati R, et al.
Synthetic Communications, 23, 3095-3095 (1993)
Reactions of methylated 5-chloro-1,2,3-thiadiazolium salts with cyclohexane-1,3-diones: X-ray crystal structure analysis of the thiapentalenic products
Gerrit L, et al.
Journal of the Chemical Society. Perkin Transactions 1, 3051-3051 (1993)
Combining click-multicomponent reaction: one-pot synthesis of triazolyl methoxy-phenyl indazolo [2, 1-b] phthalazine-trione derivatives
Salehi P, et al.
Molecular Diversity, 16(2), 231-240 (2012)
K7 [PW11CoO40]-catalyzed one-pot synthesis of polyhydroquinoline derivatives via the Hantzsch three component condensation
Heravi MM, et al.
J. Mol. Catal. A: Chem., 264(1-2), 50-52 (2007)
Clean synthesis of 1, 8-dioxo-dodecahydroxanthene derivatives catalyzed by polyaniline-p-toluenesulfonate salt in aqueous media
John A, et al.
J. Mol. Catal. A: Chem., 248(1-2), 121-125 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service