Skip to Content
Merck
All Photos(1)

Key Documents

168653

Sigma-Aldrich

2-(Methylthio)benzothiazole

97%

Synonym(s):

2-(Methylmercapto)benzothiazole, 2-(Methylthio)benzo[d]thiazole, 2-Methylsulfanyl-1,3-benzothiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7NS2
CAS Number:
Molecular Weight:
181.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

43-46 °C (lit.)

SMILES string

CSc1nc2ccccc2s1

InChI

1S/C8H7NS2/c1-10-8-9-6-4-2-3-5-7(6)11-8/h2-5H,1H3

InChI key

UTBVIMLZIRIFFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(Methylthio)benzothiazole is the degradation product of 2-(thiocyanomethylthio)benzothiazole, a biocide used in the leather, pulp and paper industries.

Application

2-(Methylthio)benzothiazole was used in trace determination of polar 1H-benzotriazoles and benzothiazoles in drinking and surface water by liquid chromatography-electrospray mass spectrometry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fulvic-acid-enhanced biodegradation of aquatic contaminants.
D Liu et al.
Bulletin of environmental contamination and toxicology, 31(2), 203-207 (1983-08-01)
Anna Latorre et al.
Environmental pollution (Barking, Essex : 1987), 147(3), 648-655 (2006-11-23)
The efficiency of several lab scale treatments (aerobic, anaerobic and ozone or combination of these) was evaluated using two packaging board mill whitewaters. The effect of the different treatments on the elimination of the organic load, the chemical oxygen demand
Cathrin Veenaas et al.
Analytical and bioanalytical chemistry, 409(20), 4867-4883 (2017-06-25)
To investigate the wide range of pollutants occurring in sewage sludge, an analytical method for comprehensive non-target screening is needed. To the best of our knowledge, no procedures currently exist for the full screening of organic contaminants in sewage sludge
G L Larsen et al.
Xenobiotica; the fate of foreign compounds in biological systems, 18(3), 313-322 (1988-03-01)
1. When 2-methylthiobenzothiazole and 35S-labelled glutathione (GSH) were incubated with rat liver homogenates, both 35S-labelled S-(2-benzothiazolyl)-glutathione and 2-mercaptobenzothiazole were isolated from the reaction mixtures. Glutathione-S-transferase appears to be involved in the S-(2-benzothiazolyl)glutathione (GBZ) formation. 2. The evidence indicates that 2-methylthiobenzothiazole
S T Nawrocki et al.
Archives of environmental contamination and toxicology, 48(3), 344-350 (2005-03-08)
2-(Thiocyanomethylthio)benzothiazole (TCMTB) is a biocide used in the leather, pulp and paper, and water-treatment industries. TCMTB may enter aquatic ecosystems during its manufacture and use. TCMTB is environmentally unstable; therefore, it is important to evaluate the toxicity of the more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service