Skip to Content
Merck
All Photos(1)

Documents

101338

Sigma-Aldrich

Benzothiazole

96%

Synonym(s):

1,3-Benzothiazole, Benzosulfonazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NS
CAS Number:
Molecular Weight:
135.19
Beilstein:
109468
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.66 (vs air)

Quality Level

vapor pressure

34 mmHg ( 131 °C)

Assay

96%

refractive index

n20/D 1.642 (lit.)

bp

231 °C (lit.)

mp

2 °C (lit.)

density

1.238 g/mL at 25 °C (lit.)

SMILES string

c1ccc2scnc2c1

InChI

1S/C7H5NS/c1-2-4-7-6(3-1)8-5-9-7/h1-5H

InChI key

IOJUPLGTWVMSFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Interesting carbonyl equivalent. Reacts with aldehydes or ketones to generate α-hydroxy carbonyl compounds.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

224.6 °F - Pensky-Martens closed cup

Flash Point(C)

107 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bulletin of the Chemical Society of Japan, 61, 3637-3637 (1988)
Ruiqing Ni et al.
Brain : a journal of neurology, 136(Pt 7), 2217-2227 (2013-06-13)
Imaging fibrillar amyloid-β deposition in the human brain in vivo by positron emission tomography has improved our understanding of the time course of amyloid-β pathology in Alzheimer's disease. The most widely used amyloid-β imaging tracer so far is (11)C-Pittsburgh compound
Thanh Binh Nguyen et al.
Angewandte Chemie (International ed. in English), 53(50), 13808-13812 (2014-10-10)
The disproportionation of elemental sulfur at moderate temperatures is investigated in the redox condensation involving o-halonitrobenzenes 1 and benzylamines 2. As a redox moderator, elemental sulfur plays the dual role of both electron donor and acceptor, generating its lowest and
Mohd Zaheen Hassan et al.
European journal of medicinal chemistry, 58, 206-213 (2012-11-06)
A series of N-(substituted benzothiazol-2-yl)amide derivatives 2a-h and 4a-h were synthesized by the EDC coupling reactions of substituted-benzothiazol-2-amine with 4-oxo-4-phenylbutanoic acid/2-benzoyl benzoic acid and evaluated for their anticonvulsant and neuroprotective effect. N-(6-methoxybenzothiazol-2-yl)-4-oxo-4-phenylbutanamide (2f) emerged as the most effective anticonvulsant with
Ahmed Kamal et al.
Bioorganic & medicinal chemistry, 20(11), 3480-3492 (2012-05-01)
A series of chalcone-amidobenzothiazole conjugates (9a-k and 10a,b) have been synthesized and evaluated for their anticancer activity. All these compounds exhibited potent activity and the IC(50) of two potential compounds (9a and 9f) against different cancer cell lines are in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service