Skip to Content
Merck
All Photos(2)

Key Documents

D218154

Sigma-Aldrich

2,2′-Dithiobis(benzothiazole)

99%

Synonym(s):

1,2-Bis(2-benzothiazolyl) disulfide, 1,2-Bis(benzo[d]thiazol-2-yl)disulfane, 2,2′-Dibenzothiazole disulfide, 2,2′-Dibenzothiazolyl disulfide, 2,2′-Dithiobis[benzothiazole], 2-(1,3-Benzothiazol-2-yldisulfanyl)-1,3-benzothiazole, Bis(2-benzothiazolyl) disulfide, DBTD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8N2S4
CAS Number:
Molecular Weight:
332.49
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

177-180 °C (lit.)

SMILES string

S(Sc1nc2ccccc2s1)c3nc4ccccc4s3

InChI

1S/C14H8N2S4/c1-3-7-11-9(5-1)15-13(17-11)19-20-14-16-10-6-2-4-8-12(10)18-14/h1-8H

InChI key

AFZSMODLJJCVPP-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samir Chahine et al.
Chemical communications (Cambridge, England), (6)(6), 592-594 (2007-02-01)
The solid state interaction of mono and macromolecular thiols at a disulfide heterocycle is shown to provide a versatile pathway for their speciation.
An unusual case of contact allergy to mercaptobenzothiazole in antifreeze.
M Kieć-Swierczynska et al.
Contact dermatitis, 41(5), 303-304 (1999-12-20)
C Herouet et al.
In vitro & molecular toxicology, 13(2), 113-123 (2000-10-14)
Contact sensitivity is a T-cell-mediated immune disease that can occur when low-molecular-weight chemicals penetrate the skin. In vivo topical application of chemical sensitizers results in morphological modification of Langerhans cells (LC). Moreover, within 18 h, LC increase their major histocompatibility
Rosana Lazzarini et al.
Dermatitis : contact, atopic, occupational, drug, 15(3), 125-130 (2005-02-24)
Contact dermatitis of the feet is a common dermatosis that very often makes patients unable to perform daily activities. The differential diagnosis should be made with other dermatoses, such as tinea, atopic dermatitis, psoriasis, dyshidrosis, and nummular eczema. It is
C Hansson et al.
Contact dermatitis, 36(4), 195-200 (1997-04-01)
At evaluation of contact dermatitis caused by solid material, patch testing is usually performed with the material as such and with extracts of it. In this study, optimization of the extraction technique monitored by quantitative HPLC analysis of the extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service