Skip to Content
Merck
All Photos(1)

Key Documents

C99000

Sigma-Aldrich

Cycloheptanone

99%

Synonym(s):

Ketocycloheptane, Ketoheptamethylene, Suberone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H12(=O)
CAS Number:
Molecular Weight:
112.17
Beilstein:
969823
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.461 (lit.)

bp

179 °C (lit.)

density

0.951 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCC1

InChI

1S/C7H12O/c8-7-5-3-1-2-4-6-7/h1-6H2

InChI key

CGZZMOTZOONQIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F

Flash Point(C)

55 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian T Sullivan et al.
Journal of chemical ecology, 47(1), 10-27 (2021-01-07)
We investigated geographic variation in the semiochemistry of major disturbance agents of western North American pine forests, Dendroctonus brevicomis Le Conte and Dendroctonus barberi Hopkins (Coleoptera: Curculionidae: Scolytinae), species separated by the Great Basin in the USA that until recently
K D Holland et al.
Molecular pharmacology, 37(1), 98-103 (1990-01-01)
The convulsant and/or anticonvulsant activity of unsubstituted and mono-alkyl-substituted cyclopentanones and cyclohexanones were examined by testing the ability of these compounds to produce seizures or to inhibit seizures induced by pentylenetetrazol and maximal electroshock in CF-1 mice. In addition, these
Khairia M Youssef et al.
Archiv der Pharmazie, 337(1), 42-54 (2004-02-05)
New series of 3, 5-bis(substituted benzylidene)-4-piperidones, 2, 7-bis(substituted benzylidene)cycloheptanones, 1, 5-bis(substituted phenyl)-1, 4-pentadien-3-ones, 1, 7-bis(substituted phenyl)-1, 6-heptadien-3, 5-diones, 1, 1-bis(substituted cinnamoyl)-cyclopentanes, and 1, 1-bis(substituted cinnamoyl)cyclohexanes have been synthesized and tested for their antioxidant activity. Among the tested compounds, compounds II(4)
Avidor Shulman et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 8(1), 229-239 (2002-02-02)
Antibody 38C2 efficiently catalyzes deuterium-exchange reactions at the alpha position of a variety of ketones and aldehydes, including substrates that have a variety of sensitive functional groups. In addition to the regio- and chemoselectivity of these reactions, the catalytic rates
Redouane Beniazza et al.
The Journal of organic chemistry, 76(3), 791-799 (2011-01-13)
A short access to homocalystegine analogues silylated at C7 is described. The synthesis involves the desymmetrization of a (phenyldimethylsilyl)methylcycloheptatriene using osmium-mediated dihydroxylation, followed by the diol protection and a cycloaddition involving the remaining diene moiety and an acylnitroso reagent. Additions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service