Skip to Content
Merck
All Photos(2)

Key Documents

158011

Sigma-Aldrich

Cyclopropylbenzene

97%

Synonym(s):

Phenylcyclopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C3H5
CAS Number:
Molecular Weight:
118.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.533 (lit.)

bp

173.6 °C/753 mmHg (lit.)

density

0.94 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

C1CC1c2ccccc2

InChI

1S/C9H10/c1-2-4-8(5-3-1)9-6-7-9/h1-5,9H,6-7H2

InChI key

VFSFCYAQBIPUSL-UHFFFAOYSA-N

Related Categories

General description

Cyclopropylbenzene is a cyclopropylarene and its oxidation by rabbit liver microsomal cytochrome P-450 has been studied. Gas-phase structure of cyclopropylbenzene has been studied by ab initio computational, microwave spectroscopic and electron diffraction techniques.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Suckling et al.
The Biochemical journal, 232(1), 199-203 (1985-11-15)
The arylcyclopropanes (cyclopropylarenes) cyclopropylbenzene and diphenylcyclopropane are oxidized by rabbit liver microsomal cytochrome P-450, both by the microsomal fraction and by the purified cytochrome in a reconstituted system. The products formed, principally benzoic acid, are due to an unusual triple
Q Shen et al.
The Journal of organic chemistry, 66(17), 5840-5845 (2001-08-21)
Ab initio computational, microwave spectroscopic, and electron diffraction techniques have been used to study the gas-phase structure of cyclopropylbenzene. Theoretical calculations at the HF, B3LYP, and MP2 levels for basis sets 6-31G(d) and 6-311G(d) have been carried out. Both MP2
The oxidation of cyclopropyl benzene by rat liver microsomal cytochrome P-450: an unusual triple oxidation of a substrate.
K E Suckling et al.
FEBS letters, 145(2), 179-181 (1982-08-23)
P Riley et al.
Xenobiotica; the fate of foreign compounds in biological systems, 24(1), 1-16 (1994-01-01)
1. The metabolism of cyclopropylbenzene (1a) and 4-cyclopropylanisole (1b) was studied using liver microsomal preparations from control, phenobarbital- and beta-naphthoflavone treated rats. 2. With all three types of microsomes 1a was metabolized by benzylic hydroxylation to give 1-phenylcyclopropanol and by
P Taavitsainen et al.
Drug metabolism and disposition: the biological fate of chemicals, 29(3), 217-222 (2001-02-22)
Currently, there are no selective, well characterized inhibitors for CYP2A6. Therefore, the effects of trans-(+/-)-2-phenylcyclopropylamine (tranylcypromine), a potent CYP2A6 inhibitor, on human liver microsomal cytochromes P450 (CYP) were studied to elucidate its selectivity. The IC50 value of tranylcypromine in coumarin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service