Skip to Content
Merck
All Photos(2)

Key Documents

179825

Sigma-Aldrich

Borane dimethyl sulfide complex

Synonym(s):

(Dimethyl sulfide)trihydroboron, BMS, Borane-dimethyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S · BH3
CAS Number:
Molecular Weight:
75.97
Beilstein:
3663489
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

density

0.801 g/mL at 25 °C (lit.)

functional group

thioether

storage temp.

2-8°C

SMILES string

B.CSC

InChI

1S/C2H6S.BH3/c1-3-2;/h1-2H3;1H3

InChI key

RMHDLBZYPISZOI-UHFFFAOYSA-N

General description

Borane dimethyl sulfide complex (BMS) is a commonly used reagent and a mediator for hydroboration reaction for the preparation of organoborane compounds, which are utilized as key intermediates in organic synthesis. BMS is also employed as a reducing agent for the reduction of various functional groups such as aldehydes, ketones, epoxides, esters, and carboxylic acids to corresponding alcohols.

Application

Borane-dimethyl sulfide (BH3 Me2S) can be used as a reagent:
  • For the selective synthesis of 1,3,5-oxygenated compounds from dimethyl 3-oxoglutarate.
  • For the conversion of ozonides to alcohols.
  • In the CBS-catalyzed asymmetric reduction of ferrocenyl-1,3-diketones to 1,3-diols.
  • For enantioselective reduction of ketones to chiral secondary alcohols in the presence of C3-symmetric tripodal hydroxyamide as a ligand.
  • For the hydroboration reduction and other applications.
  • With a dendrimeric supported L-pyrrolidinol in the asymmetric reduction of indanones and tetralones.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Other Notes

10.0-10.2 M as BH3. May contain excess methyl sulfide.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

64.4 °F

Flash Point(C)

18 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of Hydroboration of (μ-H)2Os3(CO)10 with Various Borane Complexes(BH3?L: L=Lewis base)
Chung JH
J. Korean Chem. Soc., 47, 675-678 (2003)
Efficient synthesis of 1, 3, 5-oxygenated synthons from dimethyl 3-oxoglutarate: first use of borane-dimethyl sulfide complex as a regioselective reducing agent of 3-oxygenated glutarate derivatives
Riatto VB, et al.
Journal of the Brazilian Chemical Society, 22(1), 172-175 (2011)
Highly catalytic enantioselective reduction of aromatic ketones using chiral polymer-supported Corey, Bakshi, and Shibata catalysts
Degni S, et al.
Tetrahedron Asymmetry, 15, 1495-1499 (2004)
A convenient method for the reduction of ozonides to alcohols with borane-dimethyl sulfide complex
Flippin LA, et al.
The Journal of Organic Chemistry, 54(6), 1430-1432 (1989)
Generation of bis (pentafluorophenyl) borane-dimethyl sulfide complex as a solution of hexane and its application to hydroboration of alk-1-yne with pinacolborane
Hoshi M, et al.
Tetrahedron Letters, 48(48), 8475-8478 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service