Skip to Content
Merck
All Photos(1)

Key Documents

14814

Sigma-Aldrich

Palladium(II) chloride

anhydrous, 59-60% palladium (Pd) basis

Synonym(s):

Dichloropalladium, Palladium dichloride, Palladous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Pd
CAS Number:
Molecular Weight:
177.33
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

concentration

59-60% Pd

mp

678-680 °C (lit.)

density

4 g/mL at 25 °C (lit.)

SMILES string

Cl[Pd]Cl

InChI

1S/2ClH.Pd/h2*1H;/q;;+2/p-2

InChI key

PIBWKRNGBLPSSY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.
Palladium catalysts and precursor used as an oxidizing agent and as a source of Pd(0) complexes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Yang et al.
ChemSusChem, 13(10), 2621-2627 (2020-02-11)
Metal-CO2 batteries, an attractive technology for both energy storage and CO2 utilization, are typically classified into organic Li(Na)-CO2 batteries with a high energy density/output voltage and aqueous Zn-CO2 batteries with flexible chemical production. However, achieving both high-efficiency energy storage and
Feng-Quan Yuan et al.
Chemical communications (Cambridge, England), 47(18), 5289-5291 (2011-04-01)
We present a PdCl(2)-catalyzed protocol for highly efficient allylation and benzylation of a rich variety of N-, O-, and S-containing heteroarenes under base/acid, additive, and ligand-free conditions. The method represents the very few examples for simple, universally applicable, clean, and
[(RCN)2PdCl2]-catalyzed E/Z isomerization of alkenes: a non-hydride binuclear addition-elimination pathway.
Emily H P Tan et al.
Angewandte Chemie (International ed. in English), 50(41), 9602-9606 (2011-09-23)
Maria Elisabete Machado et al.
Journal of chromatography. A, 1274, 165-172 (2013-01-10)
The separation of the organic sulfur compounds (OSC) of petroleum or its heavy fractions is a critical step and is essential for the correct characterization of these compounds, especially due to similar physical and chemical properties of polycyclic aromatic sulfur
Chun Liu et al.
Organic & biomolecular chemistry, 9(4), 1054-1060 (2010-12-15)
A simple and efficient protocol has been developed for the PdCl(2)-catalyzed ligand-free and aerobic Suzuki reaction of aryl bromides or nitrogen-based heteroaryl bromides with arylboronic acids in good to excellent yields in aqueous ethanol. A systematic investigation on the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service