Skip to Content
Merck
All Photos(3)

Key Documents

323373

Sigma-Aldrich

Palladium(II) chloride

99.995%

Synonym(s):

Dichloropalladium, Palladium dichloride, Palladous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Pd
CAS Number:
Molecular Weight:
177.33
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.995%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

678-680 °C (lit.)

density

4 g/mL at 25 °C (lit.)

SMILES string

Cl[Pd]Cl

InChI

1S/2ClH.Pd/h2*1H;/q;;+2/p-2

InChI key

PIBWKRNGBLPSSY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Palladium(II) chloride is used as a precursor to prepare palladium catalysts for various reactions like Heck coupling, cascade reaction, Buchward-Hartwig coupling. It is also used as an oxidizing agent.

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Used in the synthesis of semiconducting metal-containing polymers in which the polypyrrole backbone has a conformational energy minimum and is nearly planar.
Palladium(II) chloride can be used as a catalyst in:
  • Carbonylation of ketones to yield diesters.
  • Homo-coupling of aryl bromides using ascorbic acid and EDTA..
  • Acetylation of alcohols with vinyl acetate.
  • Arylation of 2-furaldehyde to yield 5-aryl-2-formylfuran derivatives.

Pd precursor used as an oxidizing agent and as a source of Pd(0) complexes, e.g. Heck coupling, Cascade reaction, Buchward-Hartwig coupling.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium (II) chloride/EDTA-catalyzed biaryl homo-coupling of aryl halides in aqueous medium in the presence of ascorbic acid
Ram RN and Singh V
Tetrahedron Letters, 47(43), 7625-7628 (2006)
Palladium(II) chloride
e-EROS Encyclopedia of Reagents for Organic Synthesis (2004)
Palladium (II) chloride catalyzed selective acetylation of alcohols with vinyl acetate
Bosco JWJ and Saikia AK
Chemical Communications (Cambridge, England), 47(9), 1116-1117 (2004)
Regioselective palladium-catalyzed arylation of 2-furaldehyde
McClure MS, et al.
Organic Letters, 3(11), 1677-1680 (2001)
Mihai S Viciu et al.
Organic letters, 5(9), 1479-1482 (2003-04-26)
Palladacycle dimers possessing bridging halides can be easily cleaved by using N-heterocyclic carbenes (NHCs) to generate novel monomeric complexes. The structure of one of these was determined by single-crystal diffraction study and consists of a square-planar coordination around the palladium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service