Skip to Content
Merck
All Photos(1)

Key Documents

111244

Sigma-Aldrich

trans-3-Octene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CHC2H5
CAS Number:
Molecular Weight:
112.21
Beilstein:
1719386
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010221
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

33 mmHg ( 37.7 °C)

Quality Level

Assay

98%

refractive index

n20/D 1.413 (lit.)

bp

121-122 °C (lit.)

density

0.716 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CC)=C(\[H])CCCC

InChI

1S/C8H16/c1-3-5-7-8-6-4-2/h5,7H,3-4,6,8H2,1-2H3/b7-5+

InChI key

YCTDZYMMFQCTEO-FNORWQNLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-3-Octene has been used to study the effect of double bond position and chain length in a C8 alkene in oligomerization conversion.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of liquid hydrocarbon transportation fuels by oligomerization of biomass-derived C9 alkenes.
Alonso DM, et al.
Green Chemistry, 12(6), 992-999 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service