Skip to Content
Merck
All Photos(1)

Key Documents

111236

Sigma-Aldrich

trans-2-Octene

97%

Synonym(s):

β-trans-Octene, (2E)-2-Octene, (E)-2-Octene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH3
CAS Number:
Molecular Weight:
112.21
Beilstein:
1719497
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010220
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

31 mmHg ( 37.7 °C)

Quality Level

Assay

97%

refractive index

n20/D 1.413 (lit.)

bp

123 °C (lit.)

density

0.718 g/mL at 25 °C (lit.)

SMILES string

CCCCC\C=C\C

InChI

1S/C8H16/c1-3-5-7-8-6-4-2/h3,5H,4,6-8H2,1-2H3/b5-3+

InChI key

ILPBINAXDRFYPL-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-2-Octene undergoes epoxidation with two manganese(III) porphyrin complexes by meta-chloroperbenzoic acid under catalytic reaction conditions in various solvents.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Young Hyun et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(5), 1810-1818 (2012-11-28)
The participation of multiple active oxidants generated from the reactions of two manganese(III) porphyrin complexes containing electron-withdrawing and -donating substituents with peroxyphenylacetic acid (PPAA) as a mechanistic probe was studied by carrying out catalytic oxidations of cyclohexene, 1-octene, and ethylbenzene
Aleksandr Fridlyand et al.
The journal of physical chemistry. A, 119(28), 7559-7577 (2015-02-25)
The high pressure and temperature oxidation of methyl trans-2-nonenoate, methyl trans-3-nonenoate, 1-octene, and trans-2-octene are investigated experimentally to probe the influence of the double bond position on the chemical kinetics of long esters and alkenes. Single pulse shock tube experiments

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service