Skip to Content
Merck
All Photos(1)

Key Documents

110485

Sigma-Aldrich

trans-5-Decene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH(CH2)3CH3
CAS Number:
Molecular Weight:
140.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.424 (lit.)

bp

170 °C/750 mmHg (lit.)

density

0.74 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CCCC)=C(\[H])CCCC

InChI

1S/C10H20/c1-3-5-7-9-10-8-6-4-2/h9-10H,3-8H2,1-2H3/b10-9+

InChI key

UURSXESKOOOTOV-MDZDMXLPSA-N

Application

trans-5-Decene has been used to characterize the effect of cyclization and size on the stability of Criegee intermediates formed in ozonolysis. It has been added to phenoxathiin cation radical in acetonitrile solution to form bis(10-phenoxathiiniumyl) alkane adducts.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Keisean A J M Stevenson et al.
Journal of separation science, 42(11), 2013-2022 (2019-04-10)
Thermodynamics-based models have been demonstrated to be useful for predicting retention time and peak widths in gas chromatography and two-dimensional gas chromatography separations. However, the collection of data to train the models can be time consuming, which lessens the practical
Greg T Drozd et al.
The journal of physical chemistry. A, 115(17), 4381-4387 (2011-04-12)
Ozonolysis is a key reaction in atmospheric chemistry, although important details of the behavior of the ozonolysis intermediates are not known. The key intermediate in ozonolysis, the Criegee intermeiate (CI), is known to quickly isomerize, with the favored unimolecular pathway
Bing-Jun Zhao et al.
The Journal of organic chemistry, 72(16), 6154-6161 (2007-07-03)
Addition of phenoxathiin cation radical (PO*+) to acyclic alkenes in acetonitrile (MeCN) solution occurred stereospecifically to form bis(10-phenoxathiiniumyl)alkane adducts. Stereospecific trans addition is ascribed to the intermediacy of an episulfonium cation radical. The alkenes used were cis- and trans-2-butene, cis-

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service