Skip to Content
Merck
All Photos(1)

Documents

O2501

Sigma-Aldrich

1,7-Octadiene

98%

Synonym(s):

α,ψ-Octadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)4CH=CH2
CAS Number:
Molecular Weight:
110.20
Beilstein:
605288
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.422 (lit.)

bp

114-121 °C (lit.)

density

0.746 g/mL at 25 °C (lit.)

SMILES string

C=CCCCCC=C

InChI

1S/C8H14/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-8H2

InChI key

XWJBRBSPAODJER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,7-Octadiene can serve as a crosslinker and source of ethylene for a variant of Mori′s conditions in CEYM-related reactions.

Application

1,7-Octadiene has been used in a study to assess the structure and reaction rate in olefin ring-closing metathesis of a series of simple dienes. It has also been used in a study to investigate micropatterned surfaces prepared by plasma polymerization.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ramiasa-MacGregor et al.
Nanoscale, 8(8), 4635-4642 (2016-02-09)
The wetting of a material can be tuned by changing the roughness on its surface. Recent advances in the field of nanotechnology open exciting opportunities to control macroscopic wetting behaviour. Yet, the benchmark theories used to describe the wettability of
B F P McVey et al.
Langmuir : the ACS journal of surfaces and colloids, 33(35), 8790-8798 (2017-05-30)
The role surface capping molecules play in dictating the optical properties of semiconductor nanocrystals (NCs) is becoming increasingly evident. In this paper the role of surface capping molecule polarity on the optical properties of germanium NCs (Ge NCs) is explored.
Santos Fustero et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(35), 10991-10997 (2012-08-02)
The use of 1,7-octadiene as an in situ source of ethylene led us to develop a novel multicomponent tandem cross-enyne metathesis (CEYM)-Diels-Alder reaction. The process can be considered a relay metathesis, in which the ethylene liberated in the ring-closing metathesis
E Filová et al.
Physiological research, 58(5), 669-684 (2008-12-20)
Micropatterned surfaces have been used as a tool for controlling the extent and strength of cell adhesion, the direction of cell growth and the spatial distribution of cells. In this study, chemically micropatterned surfaces were prepared by successive plasma polymerization
Ian W Ashworth et al.
Chemical communications (Cambridge, England), 46(38), 7145-7147 (2010-09-08)
In the RCM reactions of a series of simple α,ω-dienes, the relative order of reactivity has been unambiguously determined showing that cyclohexene forms faster than cyclopentene or cycloheptene. 1,5-Hexadiene inhibits the RCM of 1,7-octadiene; 1,5-hexadiene cannot progress to the RCM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service