Merck
  • Occurrence and risk assessment of selected phthalates in drinking water from waterworks in China.
Occurrence and risk assessment of selected phthalates in drinking water from waterworks in China.
Environmental science and pollution research international (2015-03-11)
Xiaowei Liu, Jianghong Shi, Ting Bo, Huiyuan Li, John C Crittenden
ABSTRACT
The first nationwide survey of six phthalates (diethyl phthalate (DEP); dimethyl phthalate (DMP); di-n-butyl phthalate (DBP); butyl benzyl phthalate (BBP); bis(2-ethylhexyl) phthalate (DEHP); din-octyl phthalate (DnOP)) in drinking waters from waterworks was conducted across seven geographical zones in China. Of the six target phthalates, DBP and DEHP were the highest abundant phthalates with median (± interquartile range) values of 0.18 ± 0.47 and 0.18 ± 0.97 μg/L, respectively, but did not exceed the limit values in China's Standards for Drinking Water Quality. These phthalates in drinking water were generally higher in the northern regions of China than those in the southern and eastern regions. Based on the investigated concentrations, lifetime exposure risk assessment indicated that phthalates in drinking water did not pose carcinogenic and noncarcinogenic risks to Chinese residents, even under the conservative scenario (with 95th percentile risk). In addition, we found that DEHP contributed the greatest risk to the total exposure risk of all the selected phthalates and oral ingestion was the main exposure route for phthalates in drinking water.
Sigma-Aldrich
Diethyl ether, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Diethyl ether, for residue analysis, JIS 5000
Sigma-Aldrich
Diethyl ether, ≥99.5%
Sigma-Aldrich
Dibutyl phthalate, SAJ special grade, ≥98.0%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Diethyl ether, SAJ first grade, ≥99.0%
Sigma-Aldrich
Diethyl ether, JIS special grade, ≥99.5%
Sigma-Aldrich
Diethyl ether, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Bis(2-ethylhexyl)phthalate-3,4,5,6-d4, 98 atom % D
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Dess-Martin periodinane, 97%
Sigma-Aldrich
Bis(2-ethylhexyl) phthalate, SAJ first grade, ≥98.0%
Sigma-Aldrich
DBP, 98% (HPLC)
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, SAJ first grade, ≥98.0
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%