Skip to Content
Merck
All Photos(1)

Key Documents

40718

Sigma-Aldrich

Diethyl pyrocarbonate

96% (NT)

Synonym(s):

DEP, DEPC, Diethyl dicarbonate, Diethyl oxydiformate, Ethoxyformic acid anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(COOC2H5)2
CAS Number:
Molecular Weight:
162.14
Beilstein:
637031
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96% (NT)

form

liquid

color

APHA: ≤20

refractive index

n20/D 1.398 (lit.)
n20/D 1.398

bp

93-94 °C/18 mmHg (lit.)

solubility

95% ethanol: 4.5 g/10 mL, clear, colorless

density

1.121 g/mL at 20 °C
1.101 g/mL at 25 °C (lit.)

functional group

carbonate

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CCOC(=O)OC(=O)OCC

InChI

1S/C6H10O5/c1-3-9-5(7)11-6(8)10-4-2/h3-4H2,1-2H3

InChI key

FFYPMLJYZAEMQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl pyrocarbonate (DEP) is a bactericidal agent and decomposes in water to CO2 and ethanol. DEP inhibits enzymes such as DNase and RNase without affecting nucleic acids. It is useful for the preparation of mRNA from tissues.

Application

Diethyl pyrocarbonate may be used in the following studies:
  • For the modification of histidyl residues in proteins.
  • As a nuclease inhibitor, for the extraction of undegraded nucleic acids from etiolated and green plant tissues.
  • For the modification of linear and supercoiled plasmid DNAs.
  • As a chemical probe to investigate the secondary structure in negatively supercoiled DNA.
  • For cabethoxylation of histidine residues of actin.
Modification reagent for His and Tyr residues in proteins. Robust probe for structural disruptions in dsDNA, reacting with fully or partially unstacked bases.

Biochem/physiol Actions

Inactivates RNase in solution at about 0.1% (v/v), thus protecting RNA against degradation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

W Herr
Proceedings of the National Academy of Sciences of the United States of America, 82(23), 8009-8013 (1985-12-01)
Purine residues located within regions of DNA that have the potential to form left-handed Z-helical structures are modified preferentially by diethyl pyrocarbonate; this hyperreactivity is dependent on the degree of negative superhelicity of the circular DNA molecules. As negative superhelical
Effects of diethyl pyrocarbonate and methyl methanesulfonate on nucleic acids and nucleases.
I Fedorcsák et al.
Acta chemica Scandinavica, 20(1), 107-112 (1966-01-01)
Modification of histidyl residues in proteins by diethylpyrocarbonate.
E W Miles
Methods in enzymology, 47, 431-442 (1977-01-01)
Selective carbethoxylation of the histidine residues of actin by diethylpyrocarbonate.
G Hegyi et al.
European journal of biochemistry, 44(1), 7-12 (1974-05-02)
A new method based on the use of diethyl pyrocarbonate as a nuclease inhibitor for the extraction of undegraded nucleic acid from plant tissues.
F Solymosy et al.
European journal of biochemistry, 5(4), 520-527 (1968-09-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service