19-2420
Methanol
≥99.5%, SAJ first grade
Synonym(s):
Methyl alcohol
About This Item
97.68 mmHg ( 20 °C)
Recommended Products
Product Name
Methanol, SAJ first grade, ≥99.5%
grade
SAJ first grade
vapor density
1.11 (vs air)
vapor pressure
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)
Assay
≥99.5%
form
liquid
autoignition temp.
725 °F
expl. lim.
36 %
availability
available only in Japan
dilution
(for analytical testing)
refractive index
n20/D 1.329 (lit.)
bp
64.7 °C (lit.)
mp
−98 °C (lit.)
density
0.791 g/mL at 25 °C (lit.)
format
neat
SMILES string
CO
InChI
1S/CH4O/c1-2/h2H,1H3
InChI key
OKKJLVBELUTLKV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Comparison of aqueous and non-aqueous capillary electrophoresis for the determination of four benzalkonium chloride homologues in compound chemical disinfectants.: This study evaluates different capillary electrophoresis methods using methanol as a non-aqueous solvent, underscoring its effectiveness in achieving higher resolution and sensitivity in the analysis of complex mixtures (Ding X et al., 2024).
- Exploring the cytotoxic potential of biflavones of Araucaria cunninghamii: Precise identification combined by LC-HRMS-metabolomics and database mining, targeted isolation, network pharmacology, in vitro cytotoxicity, and docking studies.: This comprehensive study utilizes methanol in the extraction and LC-HRMS analysis of biflavones, integrating advanced analytical techniques to explore their therapeutic potentials (Jain SK et al., 2024).
- Gas chromatography mass spectrometer determination of dimethylamine impurity in N,N-dimethylformamide solvent by derivatization of N,N-dimethylbenzamide with benzoyl chloride agent.: Demonstrates the utility of methanol in enhancing the detection of volatile organic compounds through derivatization, improving analytical accuracies in industrial chemical analysis (Mondal T et al., 2024).
- Novel Approaches for the Analysis and Isolation of Benzylisoquinoline Alkaloids in Chelidonium majus.: This research uses methanol in novel analytical methodologies to isolate and analyze alkaloids, showcasing its importance in natural product research and pharmacognosy (Ganzera M et al., 2024).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
53.6 °F
Flash Point(C)
12 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service