Skip to Content
Merck
All Photos(1)

Key Documents

675393

Sigma-Aldrich

Hexane

≥95%, HPLC Plus, for residue analysis, suitable for gas chromatography (GC) and HPLC, suitable for EPA ACB B21-02, EPA OTM-45, and EPA ACB B23-05b

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC Plus
for residue analysis
Assay:
≥95%
technique(s):
HPLC: suitable
gas chromatography (GC): suitable
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%

grade

HPLC Plus
for residue analysis

Quality Level

Agency

suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Assay

≥95%

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.01% water
water

refractive index

n20/D 1.375 (lit.)

bp

69 °C (lit.)

mp

−95 °C (lit.)

solubility

ethanol: soluble(lit.)

density

0.659 g/mL at 25 °C (lit.)

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexane (n-hexane) is a volatile organic compound.[1] It can be obtained from petroleum by distillation or by electrolyzing potassium butyrate solution. It is a widely employed organic solvent for the dissolution and isolation of fats and oils.[2] Its thermodynamic properties[3] and kinetics of thermal decomposition have been studied over a range of temperature.[4] A study on the zeolite supported platinum catalyzed isomerization and aromatization of n-hexane has been reported.[5] Its removal from hexane contaminated air streams using vapor phase biological reactor (VPBR) immobilized with hexane-degrading Aspergillus niger strain has been studied.[1] The metabolism of n-hexane forms 2,5-hexanedione (2,5-HD), which is reported to induce neurotoxicity.[6]

Application

Hexane may be used in the following processes:
  • Synthesis of graft-copolymer chains of poly-(dimethylsiloxane)-g-poly(methyl methacrylate).[7]
  • Gas chromatography/mass spectrometry (GC/MS) characterization of essential oils isolated from various origins.[8]
  • To extract free and total gossypol in combination with acetone from cottonseed flakes.[9]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Covalent binding of a neurotoxic n-hexane metabolite: conversion of primary amines to substituted pyrrole adducts by 2,5-hexanedione.
A P DeCaprio et al.
Toxicology and applied pharmacology, 65(3), 440-450 (1982-09-30)
Reforming of hexane with Pt/zeolite catalysts.
Dai LX, et al.
Catalysis Letters, 45(1-2), 107-112 (1997)
Cottonseed extraction with mixtures of acetone and hexane.
Kuk MS, et al.
Journal of the American Oil Chemists' Society, 82(8), 609-612 (2005)
The thermal decomposition of n-hexane.
Ebert KH, et al.
International Journal of Chemical Kinetics, 15(5), 475-502 (1983)
Thermodynamic properties of n-hexane.
Grigoryev BA, et al.
International Journal of Thermophysics, 9(3), 439-452 (1988)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service