Merck
  • Preparation of intravenous injection nanoformulation of VESylated gemcitabine by co-assembly with TPGS and its anti-tumor activity in pancreatic tumor-bearing mice.
Preparation of intravenous injection nanoformulation of VESylated gemcitabine by co-assembly with TPGS and its anti-tumor activity in pancreatic tumor-bearing mice.
International journal of pharmaceutics (2015-09-28)
Yanyun Xu, Haijing Meng, Fang Du, Wei Lu, Shiyuan Liu, Jin Huang, Jiahui Yu
ABSTRACT
Our recent publication showed that VES-dFdC nanocapsules in pure water could be obtained via the self-assembling of VES-dFdC prodrug synthesized by coupling gemcitabine (dFdC) with vitamin E succinate (VES). To prepare the intravenous injection nanoformulation, we present here a novel strategy to improve the stability and drug concentration of VES-dFdC nanoformulation in PBS or isotonic solution. Particularly, D-α-tocopheryl polyethylene glycol succinate (TPGS), usually used as drug solubilizer and coincidentally contains the same VES moiety as VES-dFdC prodrug and PEG chain, is selected to co-assemble with VES-dFdC prodrug. The zeta potentials of all the TPGS/VES-dFdC co-assemblies were close to 0 mV, and their particle size measured by dynamic light scattering (DLS) decreased from 113 to 36 nm with increasing TPGS/VES-dFdC molar ratios from 0.15 to 1.5. Stable colloidal suspensions were obtained without aggregates in PBS at 4 °C in one month or isotonic solution at 37 °C in one week, and the weight concentration of VES-dFdC prodrug increased from 7 to 17 mg/mL when the molar ratios of TPGS/VES-dFdC ranged from 0.5/1 to 1.5/1. The concentration of VES-dFdC prodrug was high enough to be used as intravenous injection nanoformulation in nude mice. Interestingly, along with the increase of TPGS/VES-dFdC molar ratios from 0.3/1 to 1.5/1, the morphology of TPGS/VES-dFdC co-assemblies changed from loose nanocapsule to compact micelle revealed by transmission electron microscope (TEM). Finally, the co-assembly of TPGS/VES-dFdC (TPGS/VES-dFdC: 1/1) was selected as intravenous injection nanoformulation to evaluate the antitumor activity. Compared with native dFdC, TPGS/VES-dFdC nanoformulation with 0.2mmol/kg of dosage showed similar low toxicity in vivo, but 4.7 times high of tumor inhibition rate in nude mice with pre-established BxPC-3 tumors.
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
D-α-Tocopherol succinate, analytical standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
D-α-Tocopherol succinate, semisynthetic, 1210 IU/g
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
D-α-Tocopherol succinate, BioXtra, ≥98.0% (HPLC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free