Skip to Content
Merck
All Photos(2)

Key Documents

D4551

Sigma-Aldrich

N,N-Dimethylformamide

for molecular biology, ≥99%

Synonym(s):

DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

Quality Level

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

833 °F

expl. lim.

15.2 %

impurities

≤0.5% water
water

refractive index

n20/D 1.430 (lit.)

bp

153 °C (lit.)

mp

−61 °C (lit.)

density

0.944 g/mL (lit.)

suitability

suitable for solvent for chromogenic substrates (X-Gal, etc.) used in molecular biology applications.

storage temp.

room temp

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N, N-Dimethylformamide (DMF) is a solvent with a low evaporation rate, useful for preparing solutions with a variety of hydrophobic organic compounds used in molecular biology applications.

Application

N,N-Dimethylformamide was used to solubilize MTT crystals in cell viability assays. It was also used in feruloyl esterase activity assay in molds exhibiting high activity of the enzyme.
Solvent for many hydrophobic organic compounds.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren Friedman et al.
Anti-cancer drugs, 20(6), 444-449 (2009-04-23)
Curcumin, a yellow pigment and the active component of turmeric, has been shown to protect against carcinogenesis and prevent tumor development in several types of cancer. However, its low bioavailability and potency prevent it from being effective in most chemotherapeutic
M Kanauchi et al.
Journal of food science, 73(6), C458-C463 (2009-02-26)
For food industry production processes and other uses, a mold that produces high levels of feruloyl esterase was obtained from laboratory mold collections and other sources. It was Aspergillus awanmori G-2 that produces high levels of feruloyl esterase. The feruloyl
Ruiqin Wu et al.
Food microbiology, 84, 103236-103236 (2019-08-20)
Hepatitis A virus (HAV) continues to be the leading cause of viral hepatitis. HAV outbreaks have been linked to the consumption of milk, but methods for HAV detection in milk are very limited. We developed a method to concentrate HAV
Takaki Yamauchi et al.
Plant physiology, 169(1), 180-193 (2015-06-04)
In rice (Oryza sativa) roots, lysigenous aerenchyma, which is created by programmed cell death and lysis of cortical cells, is constitutively formed under aerobic conditions, and its formation is further induced under oxygen-deficient conditions. Ethylene is involved in the induction
Jixian Xu et al.
Nature communications, 6, 7081-7081 (2015-05-09)
Solution-processed planar perovskite devices are highly desirable in a wide variety of optoelectronic applications; however, they are prone to hysteresis and current instabilities. Here we report the first perovskite-PCBM hybrid solid with significantly reduced hysteresis and recombination loss achieved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service