Skip to Content
Merck
All Photos(1)

Key Documents

437573

Sigma-Aldrich

N,N-Dimethylformamide

ACS reagent, ≥99.8%

Synonym(s):

DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
grade:
ACS reagent
Assay:
≥99.8%
technique(s):
electrophoresis: suitable
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

autoignition temp.

833 °F

expl. lim.

15.2 %

technique(s)

electrophoresis: suitable

impurities

≤0.0005 meq/g Titr. acid
≤0.003 meq/g Titr. base
≤0.15% water

evapn. residue

≤0.005%

color

APHA: ≤15

refractive index

n20/D 1.430 (lit.)

pH

6.7

bp

153 °C (lit.)

mp

−61 °C (lit.)

solubility

water: miscible

density

0.944 g/mL (lit.)

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylformamide (DMF) is a polar organic solvent with a wide range of industrial applications.[1] It is a precursor for numerous reactions such as formylation, aminocarbonylation, amination, amidation and cyanation.[2] Along with phosphorus oxychloride, it forms Vilsmeier reagent used in the formylation of reactive aromatic and heteroaromatic substrates.[3] Dielectric relaxation experiments on DMF have been conducted between 238.15K and 338.15K.[4] The thermophysical properties of the molecular interactions between DMF and the ionic liquids, ammonium salts and imidazolium salts have been studied.[5] The characteristics of solute-solvent interactions between ammonium nitrate and DMF have been investigated.[6]

Application

N,N-Dimethylformamide (DMF) may be used in the following processes:
  • As a solvent synthesis of gold nanoparticles.[7][8]
  • As a solvent in capillary electrophoresis.[9]
  • Regioselective synthesis of N,N-dimethylaminopyridzines by reacting with choropyridazines.[10]
  • Synthesis of urea-N,N-dimethylformamide, a 3:1 solvate with urea.[11]
Solvent for many hydrophobic organic compounds.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of chloropyridazines with N,N -dimethylformamide.
Lee WS, et al.
Journal of Heterocyclic Chemistry, 37(6), 1591-1591 (2000)
Urea-N,N-dimethylformamide (3/1).
Fernandes P, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(12), 4861-4861 (2007)
Capillary electrophoresis in N,N-dimethylformamide.
Porras SP and Kenndler E.
Electrophoresis, 26(17), 3279-3291 (2005)
Preparation of gold nanoparticles in formamide and N,N-dimethylformamide in the presence of poly (amidoamine) dendrimers with surface methyl ester groups.
Esumi K, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 189(1), 155-161 (2001)
Review article on Vilsmeier Haack reaction and its Applications.
Beniwal M and Jain N.
European Journal of Biomedical AND Pharmaceutical sciences, 2(3), 1340-1374 (2015)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service