Merck
  • First-Principles Prediction of Enthalpies of Formation for Polycyclic Aromatic Hydrocarbons and Derivatives.
First-Principles Prediction of Enthalpies of Formation for Polycyclic Aromatic Hydrocarbons and Derivatives.
The journal of physical chemistry. A (2015-10-21)
Thomas C Allison, Donald R Burgess
ABSTRACT
In this article, the first-principles prediction of enthalpies of formation is demonstrated for 669 polycyclic aromatic hydrocarbon (PAH) compounds and a number of related functionalized molecules. It is shown that by extrapolating density functional theory calculations to a large basis set limit and then applying a group based correction scheme that good results may be obtained. Specifically, a mean unsigned deviation and root mean squared deviation from the experimental enthalpies of formation data of 5.0 and 6.4 kJ/mol, respectively, are obtained using this scheme. This computational scheme is economical to compute and straightforward to apply, while yielding results of reasonable reliability. The results are also compared for a smaller set of molecules to the predictions given by the G3B3 and G3MP2B3 variants of the Gaussian-3 model chemistry with a mean unsigned deviation and root mean squared deviation from the experimental enthalpies of formation of 4.5 and 4.8 kJ/mol, respectively.
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Benz[b]anthracene, sublimed grade, 99.99% trace metals basis
Sigma-Aldrich
Rhein
Sigma-Aldrich
Benz[b]anthracene, 98%
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)
Sigma-Aldrich
Benzene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Benzene, JIS special grade, ≥99.5%
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Benzene, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%