Skip to Content
Merck
All Photos(3)

Key Documents

319953

Sigma-Aldrich

Benzene

ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
Beilstein:
969212
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
bp:
80 °C (lit.)
vapor pressure:
166 mmHg ( 37.7 °C)
74.6 mmHg ( 20 °C)

grade

ACS reagent

Quality Level

vapor density

2.77 (vs air)

vapor pressure

166 mmHg ( 37.7 °C)
74.6 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

1043 °F

expl. lim.

8 %

dilution

(for analytical testing)

impurities

H2SO4, passes test (darkened)
thiophene, passes test (limit ∼1ppm)
≤0.005% S compounds
≤0.05% water (Karl Fischer)

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.501 (lit.)

bp

80 °C (lit.)

mp

5.5 °C (lit.)

density

0.874 g/mL at 25 °C (lit.)

SMILES string

c1ccccc1

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H

InChI key

UHOVQNZJYSORNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzene is a monocyclic aromatic compound used as a solvent in chemical reactions. It is an essential raw material for fuels, rubber and plastics products, detergents, pesticides, and pharmaceuticals.

Application

Benzene can be used as a solvent in the:
  • Heck Reaction of vinyl bromides with styrenes to synthesize functionalized 1,3-dienes.
  • Gallium-catalyzed reductive lactonization of γ-keto acids to γ-lactone derivatives.
  • Anionic polymerization of styrene and 1,3-butadiene in the presence of phosphazene as a base.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cliona M McHale et al.
Carcinogenesis, 33(2), 240-252 (2011-12-15)
Benzene causes acute myeloid leukemia and probably other hematological malignancies. As benzene also causes hematotoxicity even in workers exposed to levels below the US permissible occupational exposure limit of 1 part per million, further assessment of the health risks associated
Paula Mayorga Burrezo et al.
Journal of the American Chemical Society, 137(11), 3834-3843 (2015-03-03)
We have used Raman spectroscopy to study the molecular and electronic structures of the radical cations and dications of carbon-bridged oligo(para-phenylenevinylene)s (COPVn, n = 1-6) possessing consecutive fused pentagons and hexagons, up to 19, along with COPV derivatives having electron-donating
David Galbraith et al.
Critical reviews in toxicology, 40 Suppl 2, 1-46 (2010-10-14)
Over the last century, benzene has been a well-studied chemical, with some acute and chronic exposures being directly associated with observed hematologic effects in humans and animals. Chronic heavy exposures to benzene have also been associated with acute myelogenous leukemia
Deborah C Glass et al.
Occupational and environmental medicine, 71(4), 266-274 (2014-02-18)
Benzene exposure has been associated with increased risk of leukaemia and myelodysplastic syndrome. Existing studies are sparse for other lymphohaematopoietic cancer subtypes, such as myeloproliferative disease (MPD) and the related chronic myeloid leukaemia (CML). We pooled data from three petroleum
Audrey Smargiassi et al.
Environmental research, 132, 38-45 (2014-04-20)
The acute cardiorespiratory effects of air quality among children living in areas with considerable heavy industry have not been well investigated. We conducted a panel study of children with asthma living in proximity to an industrial complex housing two refineries

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service