Merck
  • Optical, Electrochemical and Thermoanalytical Investigations on Newly-Synthesized Perylene-3,4,9,10-Dianhydride Fluorescent Dyes.
Optical, Electrochemical and Thermoanalytical Investigations on Newly-Synthesized Perylene-3,4,9,10-Dianhydride Fluorescent Dyes.
Journal of fluorescence (2015-06-19)
Aamer Saeed, Ghulam Shabir, Muhammad Arshad
ABSTRACT
A series of dyes using tetrachloroperylene dianhydride as fluorescent chromophore was synthesized by the substitution of suitable aliphatic and alicyclic alcohols in alkaline medium, and evaluation of dyes was done for their optical, electrochemical and thermal properties. These dyes exhibited absorption maxima λmax in the range of 440-460 nm in aqueous medium due to the presence of highly-conjugated framework. Fluorescence spectra of these dyes in water showed sharp emission peaks with small bandwidths and large fluorescence rate constants, i e., 1.36 × 10(8) to 2.25 × 10(8) S(-1). Redox potential E1/2 and band gap energy Eg were observed in the range of -0.689 to -0.784 and 2.530 to 2.610 eV, respectively. Thermal stability was noticed up to 300 °C on the basis of TG and DTA findings. The structures of perylene-azo dyes were confirmed by FTIR and NMR spectroscopy. Graphical Abstract Optical, electrochemical and thermoanalytical investigations on newly-synthesized perylene-3,4,9,10-dianhydride fluorescent dyes.
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Supelco
1-Butanol, suitable for HPLC, 99.8%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Propanol, JIS special grade
Sigma-Aldrich
1-Butanol, suitable for HPLC
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1-Propanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Cyclopentanol, 99%
Sigma-Aldrich
1-Decanol, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
1-Decanol, ≥98%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Cyclohexanol, ReagentPlus®, 99%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
1-Decanol, 98%
Sigma-Aldrich
Ethanol, ≥99.5%