Skip to Content
Merck
All Photos(1)

Key Documents

150584

Sigma-Aldrich

1-Decanol

98%

Synonym(s):

n-Decyl alcohol, Alcohol C10

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9OH
CAS Number:
Molecular Weight:
158.28
Beilstein:
1735221
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.5 (vs air)

Quality Level

vapor pressure

1 mmHg ( 70 °C)
8.25 mmHg ( 100 °C)

Assay

98%

form

liquid

autoignition temp.

550 °F

refractive index

n20/D 1.437 (lit.)

bp

231 °C (lit.)

mp

5-7 °C (lit.)

solubility

H2O: slightly soluble 0.0211 g/L at 20 °C

density

0.829 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCO

InChI

1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3

InChI key

MWKFXSUHUHTGQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Decanol was used to study the thermal properties of polymer-monolithic stationary phases.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - Pensky-Martens closed cup

Flash Point(C)

95 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sam Wouters et al.
Journal of separation science, 37(1-2), 179-186 (2013-10-30)
Thermal analysis and SEM were employed to gain insights in the different stages of morphology development and the thermal properties of polymer-monolithic stationary phases. The studied system was a thermally initiated free-radical copolymerization reaction at 70°C of styrene and divinylbenzene
Christopher S Lunde et al.
Antimicrobial agents and chemotherapy, 53(8), 3375-3383 (2009-05-28)
Telavancin is an investigational lipoglycopeptide antibiotic currently being developed for the treatment of serious infections caused by gram-positive bacteria. The bactericidal action of telavancin results from a mechanism that combines the inhibition of cell wall synthesis and the disruption of
Grit Neumann et al.
Applied and environmental microbiology, 72(6), 4232-4238 (2006-06-06)
The solvent-tolerant strain Pseudomonas putida DOT-T1E was grown in batch fermentations in a 5-liter bioreactor in the presence and absence of 10% (vol/vol) of the organic solvent 1-decanol. The growth behavior and cellular energetics, such as the cellular ATP content
Maen M Husein et al.
Journal of colloid and interface science, 288(2), 457-467 (2005-06-02)
A novel method of preparing AgCl nanoparticles by mixing AgCl powder and a microemulsion consisting of dioctyldimethylammonium chloride/n-decanol/water/isooctane is introduced. This new method was discovered during the preparation of AgCl nanoparticles in single microemulsions by direct reaction with the dioctyldimethylammonium
Maria Paola Tampieri et al.
Mycopathologia, 159(3), 339-345 (2005-05-11)
Many volatile oils are known to possess antifungal properties and are potentially applicable as antimycotic agents. By studying the efficacy of essential oils against different pathogenic mycetes, we have evaluated the in-vitro inhibiting activity of some essential oils and their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service