Merck
  • Development and validation of an UHPLC-MS/MS method for the determination of mycotoxins in grass silages.
Development and validation of an UHPLC-MS/MS method for the determination of mycotoxins in grass silages.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-09-17)
Cormac McElhinney, Pádraig O'Kiely, Chris Elliott, Martin Danaher
ABSTRACT
An ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) multi-mycotoxin analytical method was developed to simultaneously identify and quantify 20 mycotoxins in grass silages, inclusive of mycotoxins that are currently regulated in European Union feeds. Extraction of mycotoxins from dried grass silages was performed using of a modified QuEChERS extraction employing an acidified aqueous extraction (0.1 N HCl) with no further clean-up. Following chromatographic separation, analytes were detected using a fast polarity-switching MS/MS method that allowed both positive and negative ions to be analysed from a single injection, thus the reducing time and cost of analysis. The limits of detection and quantification ranged between 3 µg kg(-1) DM (aflatoxin B1, beauvericin and enniatin A and A1) and 200 µg kg(-1) DM (deoxynivalenol), and between 10 µg kg(-1) DM (aflatoxin B1, beauvericin and enniatin A1) and 500 µg kg(-1) DM (deoxynivalenol), respectively. Inter-assay accuracy and precision ranged between 90% and 107% and between 3.9% and 15.0% CV, respectively. The accuracy of the method was assessed through the application to a range of incurred samples in an inter-laboratory study.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Aflatoxin G1, from Aspergillus flavus
Sigma-Aldrich
Deoxynivalenol
Sigma-Aldrich
Beauvericin, ≥97% (HPLC)
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Fumonisin B1 from Fusarium moniliforme, ≥98% (HPLC)
Sigma-Aldrich
Aflatoxin G2
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Zearalenone, fungal mycotoxin
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%