Skip to Content
Merck
All Photos(1)

Key Documents

M3409

Sigma-Aldrich

Magnesium sulfate solution

for molecular biology, 1.00 M±0.04 M

Synonym(s):

Magnesium Sulphate, Magnesium(II) sulfate, MgSO4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.31
grade:
for molecular biology
form:
liquid
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

sterility

0.2 μm filtered

form

liquid

concentration

1.00 M±0.04 M

cation traces

heavy metals (as Pb): ≤5 ppm

foreign activity

DNase, RNase, nickase and protease, none detected

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Magnesium sulfate solution has been used:
  • as a component in super optimal broth with catabolite repression (SOC) medium for culturing Escherichia coli[1]
  • as a component in transition media for culturing human umbilical vein endothelial cells (HUVEC)[2]]
  • as a component in M9 buffer for worm fixation prior to nuclei extraction[3]

Biochem/physiol Actions

Magnesium sulfate has anti-convulsant, anti-inflammatory and vasodilatory properties. Magnesium sulfate relaxes smooth muscles.[4] In acute asthma patients, use both intravenous and nebulization of magnesium sulfate as additive treatment improves pulmonary function.[4][5] Magnesium sulfate reduces preterm labor in high risk pregnant women.[4]

Preparation Note

Prepared in 18 megohm water

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method for multiplexing CRISPR/Cas9 in Saccharomyces cerevisiae using artificial target DNA sequences
Giersch RM and Finnigan GC
Bio-protocol, 7(18), 321-330 (2017)
RNA Chromatin Immunoprecipitation (RNA-ChIP) in Caenorhabditis elegans
Cecere G and Grishok A
Bio-protocol, 4(24), e1358-e1358 (2014)
The value of magnesium sulfate nebulization in treatment of acute bronchial asthma during pregnancy
Badawy MSH and Hassanin IMA
The Egyptian journal of chest diseases and tuberculosis, 63(2), 285-289 (2014)
Estrogen decreases TNF-alpha and oxidized LDL induced apoptosis in endothelial cells
Florian M and Magder S
Steroids, 73(1), 47-58 (2008)
Lex W Doyle et al.
Obstetrics and gynecology, 113(6), 1327-1333 (2009-05-23)
To systematically review rates of neurologic outcomes reported in childhood for the preterm fetus exposed to antenatal magnesium sulfate. We searched the Cochrane Pregnancy and Childbirth Group's Trials Register, CENTRAL (The Cochrane Library 2008, Issue 3), relevant references from retrieved

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service