Skip to Content
Merck
All Photos(1)

Key Documents

295426

Sigma-Aldrich

Hydrogen chloride

≥99%, ReagentPlus®

Synonym(s):

Chlorohydric acid, Hydrochloric acid, Muriatic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCl
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

Product Name

Hydrogen chloride, ReagentPlus®, ≥99%

vapor density

1.3 (vs air)
1.3 (vs air)

Quality Level

vapor pressure

613 psi ( 21.1 °C)
613 psi ( 21.1 °C)

product line

ReagentPlus®

Assay

≥99%

form

gas

bp

-85 °C ((lit.))
−85 °C (lit.)

mp

-114 °C ((lit.))
−114 °C (lit.)

density

1.2 g/mL at 25 °C ((lit.))
1.2 g/mL at 25 °C (lit.)

SMILES string

Cl

InChI

[1S/ClH/h1H]
1S/ClH/h1H

InChI key

[VEXZGXHMUGYJMC-UHFFFAOYSA-N]
VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrogen chloride (HCl) exists in gaseous form under normal conditions. It can be used either in the gaseous or solution form. HCl is generally used in the synthesis of organic, inorganic compounds, salt formation, pH control, neutralizations and surface cleaning.

Application

HCl can be used:
  • As a catalyst in the synthesis of β-amino ketone in presence of H2O-SDS system by Mannich reaction.
  • In the synthesis of polymer-layered silicate nanocomposites.

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Press. Gas Compr. Gas - Skin Corr. 1A

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Corrosion inhibition of mild steel by plant extract in dilute HCl medium
Chauhan LR and Gunasekaran G
Corrosion Science, 49(3), 1143-1161 (2007)
Polymer-layered silicate nanocomposites: preparation, properties and uses of a new class of materials
Alexandre M and Dubois P
Materials Science & Engineering. R, Reports : A Review Journal, 28(1-2), 1-63 (2000)
HCl-catalyzed stereoselective Mannich reaction in H2O-SDS system
Akiyama T, et al.
Synlett, 2005(02) (2005)
Confused HCl: hydrogen chloride or hydrochloric acid?
Perumalla, SR and Sun CC
Chemistry?A European Journal , 18(21), 6462-6464 (2012)
Charlotte Strandgren et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(8), 3193-3205 (2015-04-17)
Hutchinson-Gilford progeria syndrome (HGPS) is a rare premature aging disorder that is most commonly caused by a de novo point mutation in exon 11 of the LMNA gene, c.1824C>T, which results in an increased production of a truncated form of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service