Skip to Content
Merck
All Photos(1)

Key Documents

F1147

Sigma-Aldrich

Fumonisin B1 from Fusarium moniliforme

≥98% (HPLC)

Synonym(s):

Macrofusine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H59NO15
CAS Number:
Molecular Weight:
721.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

biological source

Fusarium sp. (Fusarium moniliforme)

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

methanol: 9.80-10.20 mg/mL, clear, colorless to light yellow

storage temp.

2-8°C

SMILES string

CCCC[C@@H](C)[C@@H](OC(=O)C[C@@H](CC(O)=O)C(O)=O)[C@H](C[C@@H](C)C[C@H](O)CCCC[C@@H](O)C[C@H](O)[C@H](C)N)OC(=O)C[C@@H](CC(O)=O)C(O)=O

InChI

1S/C34H59NO15/c1-5-6-9-20(3)32(50-31(44)17-23(34(47)48)15-29(41)42)27(49-30(43)16-22(33(45)46)14-28(39)40)13-19(2)12-24(36)10-7-8-11-25(37)18-26(38)21(4)35/h19-27,32,36-38H,5-18,35H2,1-4H3,(H,39,40)(H,41,42)(H,45,46)(H,47,48)/t19-,20+,21-,22+,23+,24+,25+,26-,27-,32+/m0/s1

InChI key

UVBUBMSSQKOIBE-DSLOAKGESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fumonisin B1 is a mycotoxin found in many Fusarium fungi, such as Fusarium moniliforme, Fusarium proliferatum, and Fusarium verticillioides. This toxin can also be found in feedstocks and food stocks, such as corn / maize, that have been infected with Fusarium.

Fumonisin B1 (FB1) shares structure similarities with sphingosine. Thus fumonisin B1 can inhibit ceramide synthase (sphingosine N-acetyltransferase), which can lead to disruption of ceramide biosynthesis and complex sphingolipid biosynthesis.

Application

Fumonisin B1 from Fusarium moniliforme has been used:
  • as a standard in the analysis of mycotoxins in caecal content obtained from adult pigs
  • in the study of the effect of fumonisin B1 on the metabolism of prion protein (PrP) isoforms and the synthesis of scrapie prion protein PrPSc and
  • in the study of the toxic effects of fumonisin B1 on explants obtained from pig jejunum.

Biochem/physiol Actions

Fumonisin B1 acts as a hepatocarcinogen and causes hepatotoxicity in rats. In horses, it causes leukoencephalitis, and in pigs, it causes pulmonary edema syndrome. In China and southern parts of Africa, it is linked with high incidence of esophageal cancer in humans. It acts as an inhibitor of sphingosine N-acyltransferase enzyme (ceramide synthase), related to structural similarities between fumonisin B1 and sphingoid bases like sphingosine.
Fungal metabolite believed to cause leukoencephalomalacia in horses.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Kidney,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karina Basso et al.
Toxins, 5(12), 2341-2352 (2013-11-30)
Fusariotoxins such as fumonisin B1 (FB1) and deoxynivalenol (DON) cause deleterious effects on the intestine of pigs. The aim of this study was to evaluate the effect of these mycotoxins, alone and in combination, on jejunal explants from piglets, using
Judit Fodor et al.
Food additives and contaminants, 24(4), 416-420 (2007-04-25)
There is a lack of information on the effect of swine caecal microbiota on fumonisin metabolism. In this in vitro study, the biotransformation of fumonisin B(1) (FB(1)) by the gut microbiota of adult, healthy pigs was examined. Suspensions of caecal
Inhibition of Protein Serine/Threonine Phosphatases by Fumonisin B1, a Mycotoxin.
Fukuda H
Biochemical and Biophysical Research Communications, 220, 160-165 (1996)
Pranitha Dawlal et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 36(2), 296-307 (2019-01-25)
Consumption of fumonisin-contaminated foods has a negative influence on the health of humans (carcinogen; oesophageal cancer in Eastern Cape in South Africa). Lactic acid bacteria (LAB) have emerged as a promising natural detoxification agent against mycotoxins. The aim of this
Ronald T Riley et al.
Molecular nutrition & food research, 56(9), 1445-1455 (2012-07-21)
Fumonisins (FB) are mycotoxins found in maize. The purpose of this study was to (i) determine the relationship between FB(1) , FB(2) , and FB(3) intake and urinary excretion in humans, (ii) validate a method to isolate urinary FB on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service