Merck
  • Preparation of Highly Monodisperse Monopatch Particles with Orthogonal Click-Type Functionalization and Biorecognition.
Preparation of Highly Monodisperse Monopatch Particles with Orthogonal Click-Type Functionalization and Biorecognition.
Small (Weinheim an der Bergstrasse, Germany) (2015-06-06)
Thomas Tigges, Daniel Hoenders, Andreas Walther
ABSTRACT
Patchy particles are next generation colloidal building blocks for self-assembly and find further use as (bio) sensors. Progress in this direction crucially depends on developing straightforward preparation pathways able to provide patchy particles with highest uniformity and integrating precise, orthogonal, and spatially localized functionalizations to mediate interaction patterns. This continues to be one of the great challenges in colloid science. Herein, a method is shown utilizing functionalized random and block copolymers as microcontact printing inks to prepare patchy particles with outstanding control over patch size and quality. The polymeric nature and tight covalent attachment of the ink prevents flow of the ink over the particle during printing. This minimizes patch broadening and yields very small and extremely uniform patches, which is especially challenging for particle sizes below 10 μm. Click-type (amine/active ester, alkyne/azide, biotin/avidin) reactions can be performed selectively on the patch or on the particle body, rendering the particles interesting for application in imaging, biomolecular detection, and as advanced precision colloid-based building blocks.
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Copper(II) sulfate, JIS first grade, ≥97.5%
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), SAJ first grade, ≥98.0%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
2-Aminoethyl methacrylate hydrochloride, contains ~500 ppm phenothiazine as stabilizer, 90%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
6-Aminofluorescein, BioReagent, suitable for fluorescence, ~95% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Copper(II) sulfate, anhydrous, powder, ≥99.99% trace metals basis