Skip to Content
Merck
All Photos(3)

Key Documents

632287

Sigma-Aldrich

VenPure® SF

powder

Synonym(s):

Sodium borohydride, Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

reagent type: reductant

mp

>300 °C (dec.) (lit.)

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

VenPure® SF (Sodium borohydride powder) is a mild reducing agent used in the reduction of aldehydes and ketones to their corresponding alcohols. It is generally inert to other functional groups such as epoxides, esters, nitriles and lactones. Along with carboxylic acid, it forms acyloxyborohydride, which is a versatile reducing agent to reduce indoles, quinolones, imines, enamines, oximes and enamides.

Legal Information

VenPure is a registered trademark of Ascensus Specialties LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium borohydride.
Banfi L, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis., 1-13 (2014)
Sodium borohydride in carboxylic acid media: a phenomenal reduction system.
Gribble G W
Chemical Society Reviews, 27(6), 395-404 (1998)
Forty years of hydride reductions.
Brown H C and Krishnamurthy S
Tetrahedron, 35(5), 567-607 (1979)
Qinghua Cui et al.
Analytica chimica acta, 724, 86-91 (2012-04-10)
Silver nanoclusters (Ag NCs) templated with DNAs have attracted much attention as novel fluorophores because of their convenient emission tunability by the sequence and length of the template DNAs. However, the precise production of Ag NCs in a site-specific manner
Meganne L Christian et al.
ACS nano, 6(9), 7739-7751 (2012-08-10)
Owing to its high storage capacity (10.8 mass %), sodium borohydride (NaBH(4)) is a promising hydrogen storage material. However, the temperature for hydrogen release is high (>500 °C), and reversibility of the release is unachievable under reasonable conditions. Herein, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service