Skip to Content
Merck
All Photos(1)

Key Documents

05-6240

Sigma-Aldrich

Copper(II) sulfate

JIS first grade, ≥97.5%

Synonym(s):

Cupric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4
CAS Number:
Molecular Weight:
159.61
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS first grade

vapor pressure

7.3 mmHg ( 25 °C)

Assay

≥97.5%

form

powder

availability

available only in Japan

mp

200 °C (dec.) (lit.)

density

3.603 g/mL at 25 °C (lit.)

SMILES string

[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

ARUVKPQLZAKDPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nelson Durán et al.
SpringerPlus, 3, 645-645 (2014-12-09)
In the present study, semi-purified laccase from Trametes versicolor was applied for the synthesis of silver nanoparticles, and the properties of the produced nanoparticles were characterized. All of the analyses of the spectra indicated silver nanoparticle formation. A complete characterization
William T Triplett et al.
Magnetic resonance in medicine, 72(1), 8-19 (2013-09-06)
The relationship between fat fractions (FFs) determined based on multiple TE, unipolar gradient echo images and (1) H magnetic resonance spectroscopy (MRS) was evaluated using different models for fat-water decomposition, signal-to-noise ratios, and excitation flip angles. A combination of single-voxel
Mari Hansi et al.
Environmental pollution (Barking, Essex : 1987), 184, 443-448 (2013-10-15)
The density-dependence of terrestrial plant-plant interactions in the presence of toxins has previously been explored using biodegradable compounds. We exposed barley and lettuce to four copper concentrations at four stand densities. We hypothesized that toxin effects would decrease and Cu
M Seder-Colomina et al.
Ecotoxicology (London, England), 22(1), 199-205 (2012-11-10)
Microbial mats are coastal ecosystems that consist mainly of cyanobacteria, primary producers in these habitats that play an important role in stabilising delta sediments. However, these ecosystems are subject to various kinds of pollution, including metal contamination, placing their survival
Hung T Vu et al.
Environmental toxicology and chemistry, 33(4), 791-797 (2013-12-10)
The assumption of the individual effective dose is the basis for the probit method used for analyzing dose or concentration-response data. According to this assumption, each individual has a uniquely innate tolerance expressed as the individual effective dose (IED) or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service