Merck
Fenretinide: a novel treatment for endometrial cancer.
PloS one (2014-10-24)
Navdha Mittal, Saurabh Malpani, Matthew Dyson, Masanori Ono, John S Coon, Julie J Kim, Julian C Schink, Serdar E Bulun, Mary Ellen Pavone
ABSTRACT
Resistance to progestin treatment is a major hurdle in the treatment of advanced and reoccurring endometrial cancer. Fenretinide is a synthetic retinoid that has been evaluated in clinical trials as a cancer therapeutic and chemo-preventive agent. Fenretinide has been established to be cytotoxic to many kinds of cancer cells. In the present study, we demonstrate that fenretinide decreased cell viability and induced apoptosis in Ishikawa cells, which are an endometrial cancer cell line, in dose dependent manner in-vitro. This effect was found to be independent of retinoic acid nuclear receptor signaling pathway. Further, we have shown that this induction of apoptosis by fenretinide may be caused by increased retinol uptake via STRA6. Silencing of STRA6 was shown to decrease apoptosis which was inhibited by knockdown of STRA6 expression in Ishikawa cells. Results of an in-vivo study demonstrated that intraperitoneal injections of fenretinide in endometrial cancer tumors (created using Ishikawa cells) in mice inhibited tumor growth effectively. Immunohistochemistry of mice tumors showed a decrease in Ki67 expression and an increase in cleaved caspase-3 staining after fenretinide treatment when compared to vehicle treated mice. Collectively, our results are the first to establish the efficacy of fenretinide as an antitumor agent for endometrial cancer both in-vitro and in-vivo, providing a valuable rationale for initiating more preclinical studies and clinical trials using fenretinide for the treatment of endometrial cancer.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
BIS-TRIS
Megestrol acetate, European Pharmacopoeia (EP) Reference Standard
SAFC
BIS-TRIS
USP
Megestrol acetate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Citric acid trisodium salt, ≥98% (GC), anhydrous
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Retinoic acid p-hydroxyanilide, ≥95%