Skip to Content
Merck
All Photos(2)

Key Documents

RES1161B-A7

SAFC

BIS-TRIS

Synonym(s):

2,2-Bis(hydroxymethyl)-2,2′,2″-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19NO5
CAS Number:
Molecular Weight:
209.24
Beilstein:
2205275
EC Number:
MDL number:
UNSPSC Code:
12161705
NACRES:
NA.21

biological source

synthetic

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, microbial, and trace metals, tested

useful pH range

5.8-7.2

pKa (25 °C)

6.5

solubility

H2O: 0.1 M, clear, colorless (aqueous)

suitability

suitable for manufacturing use

foreign activity

Cytotoxicity, DNase, NICKase, RNase, and Protease; tested

SMILES string

OCCN(CCO)C(CO)(CO)CO

InChI

1S/C8H19NO5/c10-3-1-9(2-4-11)8(5-12,6-13)7-14/h10-14H,1-7H2

InChI key

OWMVSZAMULFTJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Buffer quality is vital for the success of biopharmaceutical processes, because buffers are indispensable in nearly every production step.

Our broad portfolio of buffer materials manufactured under appropriate controls is tailored to your needs. Ranging from non-GMP grades for low-risk application, to IPEC-PQG GMP for higher-risk applications, we have products covering all your manufacturing needs.

Application

Bis Tris is a commonly used biological buffer for very broad usage in biological systems such as diagnostics, electrophoresis and protein based bioprocessing. The pKa of Bis Tris is 6.36 which makes it a good candidate for biological based formulations.

Bis Tris is used in a wide variety of biological applications including culture media, electrophoresis separations and diagnostic reagent formulations.

Packaging

Product is available in the following package sizes:
RES1161B-A701X: 100g container
RES1161B-A702X 1kg container
RES1161B-A704X: 10kg container
RES1161B-A705X: 25kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zahra Sabouri et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(25), E2567-E2575 (2014-05-14)
The best-understood mechanisms for achieving antibody self/non-self discrimination discard self-reactive antibodies before they can be tested for binding microbial antigens, potentially creating holes in the repertoire. Here we provide evidence for a complementary mechanism: retaining autoantibodies in the repertoire displayed
Alastair W Skeffington et al.
Plant physiology, 165(2), 866-879 (2014-05-02)
The first step on the pathway of starch degradation in Arabidopsis (Arabidopsis thaliana) leaves at night is the phosphorylation of starch polymers, catalyzed by glucan, water dikinase (GWD). It has been suggested that GWD is important for the control of
Elizabeth M MacDonald et al.
Disease models & mechanisms, 7(4), 471-481 (2014-02-08)
The purpose of our study was to compare two acquired muscle atrophies and the use of myostatin inhibition for their treatment. Myostatin naturally inhibits skeletal muscle growth by binding to ActRIIB, a receptor on the cell surface of myofibers. Because
Lesley Cheng et al.
Kidney international, 86(2), 433-444 (2013-12-20)
Micro RNAs (miRNAs) have been shown to circulate in biological fluids and are enclosed in vesicles such as exosomes; they are present in urine and represent a noninvasive methodology to detect biomarkers for diagnostic testing. The low abundance of RNA
X Chen et al.
Oncogene, 33(39), 4724-4734 (2013-10-22)
Uveal melanoma (UM) is a genetically and biologically distinct type of melanoma, and once metastatic there is no effective treatment currently available. Eighty percent of UMs harbor mutations in the Gαq family members GNAQ and GNA11. Understanding the effector pathways

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service