Merck
  • Multi-residue Determination of Polar Veterinary Drugs in Livestock and Fishery Products by Liquid Chromatography/Tandem Mass Spectrometry.
Multi-residue Determination of Polar Veterinary Drugs in Livestock and Fishery Products by Liquid Chromatography/Tandem Mass Spectrometry.
Journal of AOAC International (2015-04-11)
Maki Kanda, Takayuki Nakajima, Hiroshi Hayashi, Tsuneo Hashimoto, Setsuko Kanai, Chieko Nagano, Yoko Matsushima, Yukinari Tateishi, Soichi Yoshikawa, Yumi Tsuruoka, Takeo Sasamoto, Ichiro Takano
ABSTRACT
Residues of 37 polar veterinary drugs belonging to six families (quinolones, tetracyclines, macrolides, lincosamides, sulfonamides, and others) in livestock and fishery products were determined using a validated LC-MS/MS method. There were two key points in sample preparation. First, extraction was performed with two solutions of different polarity. Highly polar compounds were initially extracted with Na2EDTA-McIlvaine's buffer (pH 7.0). Medium polar compounds were then extracted from the same samples with acetonitrile containing 0.1% formic acid. Secondly, cleanup was performed using a single SPE polymer cartridge. The first extracted solution was applied to the cartridge. Highly polar compounds were retained on the cartridge. Then, the second extracted solution was applied to the same cartridge. Both highly and medium polar compounds were eluted from the cartridge. This method satisfied the guideline criteria for 37 out of 37 drugs in swine muscle, chicken muscle, bovine muscle, prawn, salmon trout, red sea bream, milk, and honey; 35 out of 37 in egg; and 34 out of 37 in flounder. The LOQ ranged from 0.1 to 5 μg/kg. Residues were detected in 24 out of 110 samples and analyzed using the validated method.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Millipore
Chlortetracycline Selective Supplement, suitable for microbiology
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Josamycin, ≥90% (HPLC)
Sigma-Aldrich
Sulfamonomethoxine
Supelco
Sulfadimethoxine, 98.0-102.0%
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Thiabendazole, suitable for plant cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Supelco
Sulfadimethoxine, VETRANAL®, analytical standard
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Oxytetracycline, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfadimethoxine for peak identification, European Pharmacopoeia (EP) Reference Standard
Marbofloxacin, European Pharmacopoeia (EP) Reference Standard