Skip to Content
Merck
All Photos(2)

Key Documents

T5535

Sigma-Aldrich

Thiabendazole

powder, suitable for plant cell culture, BioReagent

Synonym(s):

2-(4-Thiazolyl)benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7N3S
CAS Number:
Molecular Weight:
201.25
Beilstein:
611403
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

Product Name

Thiabendazole, suitable for plant cell culture

Quality Level

SMILES string

c1ccc2[nH]c(nc2c1)-c3cscn3

InChI

1S/C10H7N3S/c1-2-4-8-7(3-1)12-10(13-8)9-5-14-6-11-9/h1-6H,(H,12,13)

InChI key

WJCNZQLZVWNLKY-UHFFFAOYSA-N

Gene Information

human ... CYP2C9(1559)

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: imidazole

Application

Thiabendazole, a metal chelator and helminth-specific fumarate reductase inhibitor, is used primarily as an anthelmintic to control infections by helminth organisms. It is also used to control mold and fungal growth in plants.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.Storage class (TRGS 510): Non Combustible Solids

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alfredo Santovito et al.
Archives of toxicology, 85(6), 689-693 (2010-10-13)
Thiabendazole is a benzimidazole-derived compound widely employed in agriculture as anthelmintic and fungicide. It is also used as a post-harvest fungicide for imported citrus fruits during transport and storage, and thus, it was found at high concentration in fruits and
Petr Heneberg et al.
Scientific reports, 10(1), 7077-7077 (2020-04-29)
Neonicotinoid insecticides are associated with a decline in the diversity and distribution of bees and wasps (Hymenoptera: Aculeata). The effects of neonicotinoids on the metamorphosis of aculeates have never been addressed in detail; however, recent evidence suggests that neonicotinoids induce
Stuart Dashper et al.
Antimicrobial agents and chemotherapy, 54(3), 1311-1314 (2009-12-30)
Porphyromonas gingivalis is a major pathogen of chronic periodontitis and exists in a biofilm on the surface of the tooth root. Oxantel, a cholinergic anthelmintic and fumarate reductase inhibitor, significantly inhibited biofilm formation by P. gingivalis and disrupted established biofilms
Chun-Min Shan et al.
Cell reports, 33(13), 108561-108561 (2020-12-31)
One key aspect of epigenetic inheritance is that chromatin structures can be stably inherited through generations after the removal of the signals that establish such structures. In fission yeast, the RNA interference (RNAi) pathway is critical for the targeting of
P Fonderie et al.
Parasitology, 139(10), 1301-1308 (2012-06-22)
Studies on anthelmintic resistance in equine parasites do not include facultative parasites. Halicephalobus gingivalis is a free-living bacterivorous nematode and a known facultative parasite of horses with a strong indication of some form of tolerance to common anthelmintic drugs. This

Articles

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service