Skip to Content
Merck
All Photos(1)

Key Documents

Y0001679

Sulfadimethoxine for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sulfadimethoxine, 4-Amino-N-(2,6-dimethoxy-4-pyrimidinyl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O4S
CAS Number:
Molecular Weight:
310.33
Beilstein:
306856
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

sulfadimethoxine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC)n1

InChI

1S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)

InChI key

ZZORFUFYDOWNEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfadimethoxine for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minjoo Kim et al.
PloS one, 10(3), e0119519-e0119519 (2015-03-18)
Existing data on the association between being overweight and cardiovascular morbidity and mortality risk in adults are inconsistent. We prospectively and longitudinally investigated the effects of weight on arterial stiffness and plasma metabolites in middle-aged subjects (aged 40-55 years). A
Zhe Meng et al.
Food chemistry, 174, 597-605 (2014-12-23)
A novel UPLC-MS/MS method for quantification and confirmation of eight fluoroquinolones, five sulphonamides (SAs) and four acetyled metabolites in milk is developed. Their main fragmentation pathways were presented. The limits of quantification were in the range of 0.01-0.29 μg kg(-1)
Werner Siegmund et al.
British journal of clinical pharmacology, 79(3), 501-513 (2014-09-30)
The rare association of flupirtine with liver injury is most likely caused by reactive quinone diimines and their oxidative formation may be influenced by the activities of N-acetyltransferases (NAT) that conjugate the less toxic metabolite D13223, and by glucuronosyltransferases (UGT)
Sae Young Lee et al.
PloS one, 10(4), e0123306-e0123306 (2015-04-10)
Hypertriglyceridemia (HTG) is a risk factor for atherosclerotic cardiovascular disease (CVD). We investigated alterations in plasma metabolites associated with borderline-to-moderate HTG (triglycerides (TG) 150-500 mg/dL). Using UPLC-LTQ-Orbitrap mass spectrometry analysis, the metabolomics profiles of 111 non-diabetic and non-obese individuals with
Yoshiki Murakami et al.
Drug development and industrial pharmacy, 40(8), 1065-1071 (2013-06-21)
The hepatic metabolism of six compounds newly synthesized as retinoid X receptor agonists was characterized in rat and human liver microsomes to examine the relationship between their hepatic metabolism profiles and side chain structures, considering the interspecies difference. The compounds

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service