Merck
  • On the colonic bacterial metabolism of azo-bonded prodrugsof 5-aminosalicylic acid.
On the colonic bacterial metabolism of azo-bonded prodrugsof 5-aminosalicylic acid.
Journal of pharmaceutical sciences (2014-08-06)
Tiago Sousa, Vipul Yadav, Vanessa Zann, Anders Borde, Bertil Abrahamsson, Abdul W Basit
ABSTRACT
Azo-bonded prodrugs of 5-aminosalicylic acid (mesalazine)-sulfasalazine, balsalazide, and olsalazine, which are used in the treatment of ulcerative colitis, rely on colonic bacteria to cleave the azo bond and liberate the active drug in the large intestine. The aim of this study was to use an in vitro colonic simulator to determine the rates of metabolism of these three prodrugs in the presence of colonic bacteria, and to link the data to results obtained previously in humans. In individual fecal slurries prepared from five different donors, sulfasalazine degradation was rapid and virtually complete within 4 h, confirming the ubiquitous nature of azo-reduction between individuals. In pooled fecal slurry, the rate of degradation of sulfasalazine was faster (t1/2 , 32.8 min) than balsalazide (t1/2 , 80.9 min) and olsalazine (t1/2 , 145.1 min). These results are in agreement with data in humans, where it was found that sulfasalazine was more extensively metabolized on passage through the human colon than the other two drugs. These findings indicate that other than the azo bond itself, the broader chemical structure of the molecules play a role in the degradation of this class of compound, and highlight the utility of this in vitro model to evaluate the metabolism of drugs in the presence of colonic microbiota.
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture