Skip to Content
Merck
All Photos(1)

Key Documents

71386

Sigma-Aldrich

Sodium chloride solution

BioUltra, for molecular biology, ~5 M in H2O

Synonym(s):

Halite, NaCl, sodium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
for molecular biology
form:
liquid
solubility:
water: soluble

grade

for molecular biology

product line

BioUltra

form

liquid

concentration

~5 M in H2O

technique(s)

DNA extraction: suitable
LAMP: suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

refractive index

n20/D 1.378

pH

5.0-7.0 (25 °C)

solubility

water: soluble

density

1.186 g/mL (5 M in water)

cation traces

Al: ≤1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.02
λ: 280 nm Amax: 0.01

suitability

suitable for molecular biology

application(s)

genomic analysis

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium chloride is a vital nutrient that aids normal body functions in humans and animals. This chemical is commonly found in nature with a wide variety of biochemical applications, like intravenous fluids and density gradients. It is also used as a diluent to increase ionic strength in buffers or culture media. Sodium chloride is crucial for maintaining plasma osmolality and the extracellular fluid volume of the body. Excess NaCl leads to increased blood pressure, which has implications for cardiovascular and renal diseases. Sterile NaCl is a component of eye drop solutions. Traditionally, it has been used in high concentrations to preserve food since most bacteria cannot grow in high-salt conditions.

Application

Sodium chloride solution has been used as a component of:
  • polyethylene glycol (PEG) buffer for the purification of double-tagged products
  • NT2 buffer for the preparation of antibody-coated Protein A Sepharose (PAS) beads and immunoprecipitation of ribonucleoproteins (RNPs)
  • chilled lysis buffer for the isolation of single nuclei from mice kidney

Features and Benefits

  • Geologically stable
  • Suitable for purification, precipitation, and crystallization

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis Kim et al.
JAMA, 311(1), 45-52 (2013-11-19)
Hospital cooling improves outcome after cardiac arrest, but prehospital cooling immediately after return of spontaneous circulation may result in better outcomes. To determine whether prehospital cooling improves outcomes after resuscitation from cardiac arrest in patients with ventricular fibrillation (VF) and
Alexander W Wyatt et al.
Genome biology, 15(8), 426-426 (2014-08-27)
Genomic analyses of hundreds of prostate tumors have defined a diverse landscape of mutations and genome rearrangements, but the transcriptomic effect of this complexity is less well understood, particularly at the individual tumor level. We selected a cohort of 25
Laurent Dortet et al.
Journal of clinical microbiology, 52(7), 2359-2364 (2014-04-25)
Multidrug-resistant Acinetobacter baumannii isolates, particularly those that produce carbapenemases, are increasingly reported worldwide. The biochemically based Carba NP test, extensively validated for the detection of carbapenemase producers among Enterobacteriaceae and Pseudomonas spp., has been modified to detect carbapenemase production in
David G Guzzardi et al.
Journal of the American College of Cardiology, 66(8), 892-900 (2015-08-22)
Suspected genetic causes for extracellular matrix (ECM) dysregulation in the ascending aorta in patients with bicuspid aortic valves (BAV) have influenced strategies and thresholds for surgical resection of BAV aortopathy. Using 4-dimensional (4D) flow cardiac magnetic resonance imaging (CMR), we
Koji Tanaka et al.
Nature communications, 6, 6337-6337 (2015-02-27)
Pore-forming toxins (PFT) are water-soluble proteins that possess the remarkable ability to self-assemble on the membrane of target cells, where they form pores causing cell damage. Here, we elucidate the mechanism of action of the haemolytic protein fragaceatoxin C (FraC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service