Skip to Content
Merck
All Photos(1)

Key Documents

51280

Sigma-Aldrich

Hemin

BioXtra, from Porcine, ≥96.0% (HPLC)

Synonym(s):

Chloro(protoporphyrinato)iron(III), Chlorohemin, Chloroprotoporphyrin IX iron(III), Ferriprotoporphyrin IX chloride, Hemin(chloride)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H32ClFeN4O4
CAS Number:
Molecular Weight:
651.94
Beilstein:
5229914
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.79

biological source

Porcine

product line

BioXtra

Assay

≥96.0% (HPLC)

form

powder or crystals

technique(s)

tissue culture: suitable
transfection: suitable

storage temp.

2-8°C

SMILES string

Cc1c(CCC(O)=O)c2cc3c(CCC(O)=O)c(C)c4cc5nc(cc6c(C)c(C=C)c(cc1n2)n6[Fe](Cl)n34)c(C=C)c5C

InChI

1S/C34H34N4O4.ClH.Fe/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);1H;/q;;+3/p-3/b25-13-,26-13-,27-14-,28-15-,29-14-,30-15-,31-16-,32-16-;;

InChI key

BTIJJDXEELBZFS-HXFTUNQESA-K

Looking for similar products? Visit Product Comparison Guide

Application

Hemin is used for the following applications:
  • To study bacterial strains and growth conditions
  • Used in short-term colony assays (the culture medium consists of hemin along with other components)
  • Cell transfections
It may be used in β-hematin formation assay for analyzing the inhibitory activity of MMV (Medicines for Malaria Venture) Malaria Box compounds against the formation of β-hematin.

Biochem/physiol Actions

Hemin, the oxidized version of heme, is an iron-containing prosthetic group for a diverse group of proteins. Free heme, which can be released from hemoglobin following hemolysis, is pro-inflammatory and contributes to iron-derived reactive oxygen species. Free hemin levels may be upregulated in various pathological conditions and can contribute to various inflammatory conditions including vascular disorders, renal failure, and immune-mediated disorders.

Hemin may be used to study the expression and represson of heme oxygenase 1 (HO-1) as well as the activity of HO-1. Hematin is the "X factor" required to grown Haemophilus influenzae.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Curti et al.
Journal of immunology (Baltimore, Md. : 1950), 166(2), 848-854 (2001-01-06)
We studied cytokine-driven differentiation of primitive human CD34(+)HLA-DR(-) cells to myeloid dendritic cells (DC). Hemopoietic cells were grown in long-term cultures in the presence of various combinations of early acting cytokines such as FLT3-ligand (FLT3-L) and stem cell factor (SCF)
Christine C Chiu et al.
PloS one, 14(5), e0216753-e0216753 (2019-05-10)
The mountain pine beetle (MPB; Dendroctonus ponderosae) is a forest insect pest that attacks several different pine (Pinus) species in its native range of distribution in western North America. MPB are exposed for most of their life cycle to the
Kim Y Fong et al.
International journal for parasitology. Drugs and drug resistance, 5(3), 84-91 (2015-07-08)
The Malaria Box, assembled by the Medicines for Malaria Venture, is a set of 400 structurally diverse, commercially available compounds with demonstrated activity against blood-stage Plasmodium falciparum. The compounds are a representative subset of the 20,000 in vitro antimalarials identified from
S Abboud et al.
The Journal of biological chemistry, 275(26), 19906-19912 (2000-04-05)
We have isolated and characterized a novel iron-regulated gene that is homologous to the divalent metal transporter 1 family of metal transporters. This gene, termed metal transporter protein (mtp1), is expressed in tissues involved in body iron homeostasis including the
Eyal Golub et al.
The Analyst, 136(21), 4397-4401 (2011-09-02)
Hemin/G-quadruplex catalyzes the H(2)O(2)-mediated oxidation of Amplex Red to the fluorescent product resorufin. This process is implemented to develop hairpin nucleic acid structures for the detection of DNA, to probe the catalytic activity of glucose oxidase, to use the thrombin-aptamer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service