Merck
  • Efficient and simple approaches towards direct oxidative esterification of alcohols.
Efficient and simple approaches towards direct oxidative esterification of alcohols.
Chemistry (Weinheim an der Bergstrasse, Germany) (2014-10-07)
Ritwika Ray, Rahul Dev Jana, Mayukh Bhadra, Debabrata Maiti, Goutam Kumar Lahiri
ABSTRACT
The present article describes novel oxidative protocols for direct esterification of alcohols. The protocols involve successful demonstrations of both "cross" and "self" esterification of a wide variety of alcohols. The cross-esterification proceeds under a simple transition-metal-free condition, containing catalytic amounts of TEMPO (2,2,6,6-tetramethyl-1-piperidinyloxy)/TBAB (tetra-n-butylammonium bromide) in combination with oxone (potassium peroxo monosulfate) as the oxidant, whereas the self-esterification is achieved through simple induction of Fe(OAc)2 /dipic (dipic=2,6-pyridinedicarboxylic acid) as the active catalyst under an identical oxidizing environment.
Product Description
Supelco
2,6-Pyridinedicarboxylic acid concentrate, 0.02 M C7H5NO4 in water (0.04N), suitable for ion chromatography, eluent concentrate
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
1-Octanol, JIS special grade, ≥98.0%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
1-Octanol, SAJ first grade, ≥75.0%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
1-Octanol, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Supelco
2,6-Pyridinedicarboxylic acid, suitable for ion chromatography, ≥99.5% (T)
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2,6-Pyridinedicarboxylic acid, 99%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99%, FG
Sigma-Aldrich
3-Phenyl-1-propanol, 98%
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99.6%
Sigma-Aldrich
3-Phenyl-1-propanol, ≥98%, FCC
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
2-Ethyl-1-hexanol, puriss., ≥99.0% (GC)
Supelco
2-Ethyl-1-hexanol, analytical standard