Skip to Content
Merck
All Photos(1)

Key Documents

23-0130

Sigma-Aldrich

1-Octanol

SAJ first grade, ≥75.0%

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1697461
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
grade:
SAJ first grade
technique(s):
HPLC: suitable
bp:
196 °C (lit.)
vapor pressure:
0.14 mmHg ( 25 °C)

grade

SAJ first grade

vapor density

4.5 (vs air)

vapor pressure

0.14 mmHg ( 25 °C)

Assay

≥75.0%

form

liquid

autoignition temp.

523 °F

expl. lim.

8 % (lit.)

availability

available only in Japan

technique(s)

HPLC: suitable

refractive index

n20/D 1.429 (lit.)

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

177.8 °F

Flash Point(C)

81 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashraf S Al-Madhoun et al.
Cancer research, 64(17), 6280-6286 (2004-09-03)
Thymidine analogs containing o-carboranylalkyl groups at the 3-position were screened as potential substrates for human thymidine kinase 1 (TK1), an enzyme that is selectively expressed in a variety of rapidly proliferating cells, including tumor cells. On the basis of previous
R O Potts et al.
Pharmaceutical research, 9(5), 663-669 (1992-05-01)
Published permeability coefficient (Kp) data for the transport of a large group of compounds through mammalian epidermis were analyzed by a simple model based upon permeant size [molecular volume (MV) or molecular weight (MW)] and octanol/water partition coefficient (Koct). The
John A Arnott et al.
Expert opinion on drug discovery, 7(10), 863-875 (2012-09-21)
The role of lipophilicity in drug discovery and design is a critical one. Lipophilicity is a key physicochemical property that plays a crucial role in determining ADMET (absorption, distribution, metabolism, excretion, and toxicity) properties and the overall suitability of drug
B D Anderson et al.
The Journal of investigative dermatology, 93(2), 280-286 (1989-08-01)
The permeability coefficients (kp) of a series of methyl-substituted p-cresols were determined in human stratum corneum along with their partition coefficients (PC) between water and untreated stratum corneum, delipidized stratum corneum, octanol, and heptane. The PC values were identical in
Cristian Gobbo et al.
ACS nano, 6(12), 10684-10698 (2012-12-12)
Controlling the molecular conformation of oligomers on surfaces through noncovalent interactions symbolizes an important approach in the bottom-up patterning of surfaces with nanoscale precision. Here we report on the design, synthesis, and scanning tunneling microscopy (STM) investigation of linear oligoamides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service